Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2000

Sigma-Aldrich

Anti-LXR alpha Antibody, clone 2B7

clone 2B7, from mouse

Synonym(s):

Oxysterols receptor LXR-alpha, Liver X receptor alpha, Nuclear receptor subfamily 1 group H member 3, RLD-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$587.00

$587.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$587.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$587.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2B7, monoclonal

species reactivity

mouse, rat

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Nr1H3(22259)

Related Categories

General description

Oxysterols receptor LXR-alpha (UniProt: Q62685; also known as Liver X receptor alpha, Nuclear receptor subfamily 1 group H member 3, RLD-1) is encoded by the Nr1h3 (also known as Lxra) gene in rat. Liver X receptors (LXRs) are members of the nuclear receptor superfamily that have been implicated in lipid homeostasis and glucose metabolism. Oxysterols, oxidized derivatives of cholesterol, are ligands for LXRs. LXRs act as cholesterol sensors; when cellular oxysterols accumulate as a result of increasing concentrations of cholesterol, LXR induces the transcription of genes that protect cells from cholesterol overload. LXRs bind to consensus elements (LXR response elements, LXREs) as heterodimers with isoforms of the retinoid X receptor (RXR). LXR-alpha is an important regulator of cholesterol and bile acid metabolism. It interacts with RXR and shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. In adults it is expressed in spleen, pituitary, lung, liver, and fat and a weaker expression is observed in several other tissues. LXR-alpha contains a transactivation domain (AF-1; aa 1-94) that is required for ligand-independent transactivation function; another transactivation domain (AF-2; aa 203-445) that is required for ligand-dependent transactivation function; and a ligand binding domain (aa 213-432). (Ref.: Zhao, C., and Dahlman-Wright, K. (2010). J. Endocrinol. 204, 233-240).

Specificity

Clone 2B7 detects LXR-alpha in mouse and rat liver tissue.

Immunogen

GST-tagged full length recombinant rat LXR-alpha protein.

Application

Anti-LXR alpha, clone 2B7, Cat. No. MABS2000, is a mouse monocloanl antibody that detects Oxysterols receptor LXR-alpha and has been tested for use in Immunoprecipitation and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected LXR alpha in Immunoprecipitation applications (Tian, J., et. al. (2016). Proc Natl Acad Sci USA. 113(29):8182-7).

Western Blotting Analysis: A representative lot detected LXR alpha in Western Blotting applications (Tian, J., et. al. (2016). Proc Natl Acad Sci USA. 113(29):8182-7).
Research Category
Signaling

Quality

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: 1 ug/mL of this antibody detected LXR alpha in 10 µg of mouse liver tissue lysate.

Target description

~50 kDa observed; 50.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service