Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABS196

Sigma-Aldrich

Anti-mTOR/FRAP Antibody, clone 22C2

clone 22C2, from mouse

Synonym(s):

Serine/threonine-protein kinase mTOR, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, Mammalian target of rapamycin, mTOR, Mechanistic target of rapamycin, Rapamycin and FKBP12 target 1, Rapam

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22C2, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTOR(2475)

General description

The mammalian target of rapamycin (mTOR), also known as FRAP, RAFT, or RAPT, is a ubiquitously expressed serine/threonine kinase that belongs to the phosphoinositide 3-kinase–related kinase (PIKK) family. mTOR functions as a downstream effector in the PI3K pathway and is activated by the Akt kinase. mTOR responds to signals from growth factors and promotes protein synthesis underlying cell proliferation, by activation of the S6 kinase and inhibition of the 4E-BP protein involved in protein translation. Several studies have suggested that the proliferative functions of mTOR may contribute to the development of cancer.

Immunogen

Linear peptide corresponding to human mTOR/FRAP.

Application

Detect the mTOR/FRAP protein using this Anti-mTOR Antibody, clone 22C2, validated for use in WB & IP.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases
Western Blot Analysis: A representative lot was used by an independent laboratory in neuroblastoma cells. (Hosoi, H., et al. (1998). Molecular Pharmacology. 54:815–824.)

Immunoprecipitation Analysis: A representative lot was used by an independent laboratory in Rho30 cell extracts. (Hosoi, H., et al. (1998). Molecular Pharmacology. 54:815–824.)

Quality

Evaluated by Western Blot in Jurkat cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected mTOR/FRAP on 10 µg of Jurkat cell lysate.

Target description

~290 kDa observed

Linkage

Replaces: 04-385

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Jurkat cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Harris et al.
The Journal of biological chemistry, 286(49), 42615-42625 (2011-10-11)
Steatosis is a frequent complication of hepatitis C virus infection. In mice, this condition is recapitulated by the expression of a single viral protein, the nucleocapsid core. Core localizes to the surface of lipid droplets (LDs) in infected liver cells
Xiaohui Yang et al.
Cancer biology & therapy, 19(8), 706-714 (2018-04-04)
(-)-Guaiol, a sesquiterpene alcohol with the guaiane skeleton, has been found in many Chinese medicinal plants and been reported to comprise various guaiane natural products that are well known for their antibacterial activities. Previously, we have shown its antitumor activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service