Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1923

Sigma-Aldrich

Anti-PTK7 Antibody, clone 31G9

clone 31G9, from rat

Synonym(s):

Inactive tyrosine-protein kinase 7, Colon carcinoma kinase 4, CCK-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, Tyrosine-protein kinase-like 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

31G9, monoclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PTK7(5754)

General description

Inactive tyrosine-protein kinase 7 (UniProt: Q13308; also known as Colon carcinoma kinase 4, CCK-4, Protein-tyrosine kinase 7, Pseudo tyrosine kinase receptor 7, Tyrosine-protein kinase-like 7, PTK7) is encoded by the PTK7 (also known as CCK4) gene (Gene ID: 5754) in human. PTK7 is a single-pass type I membrane protein that lacks the catalytic activity. It is highly expressed in lung, liver, pancreas, kidney, placenta and melanocytes. It is weakly expressed in thyroid gland, ovary, brain, heart and skeletal muscle. Its expression is reported to be higher in cell lines established from normal non-tumorigenic tissues compared those from highly metastatic invasive carcinomas. Six isoforms of PTK7 have been described that are generated by alternative splicing. It is synthesized with a signal peptide (aa 1-30), which is subsequently cleaved off to generate mature form. It contains seven Ig-like-C2 type domains. It has an extracellular domain (aa31-704), a transmembrane domain (aa 705-725; and a cytoplasmic domain (aa 726-1070). PTK7 is shown to associate with receptor tyrosine kinase-like orphan receptor 2 (ROR2) to form a heterodimeric complex in mammalian cells and PTK7 and ROR2 physically and functionally interact with the non-canonical WNT5A ligand leading to JNK activation and cell movements. PTK7 is cleaved by MMP14 between Pro 621 and Leu 622 to generate an N-terminal soluble (70 kDa) fragment and a membrane C-terminal (50 kDa) fragment. (Ref.: Martinez, S., et al. (2015). J. Biol. Chem. 290(51); 30562-72).

Specificity

Clone 31G9 is a rat monoclonal antibody that detects human Inactive tyrosine protein kinase 7 (PTK7). It targets an epitope within the extracellular domain.

Immunogen

Epitope: extracellular domain
GST-tagged recombinant fragment corresponding to the extracellullar domain of human Inactive tyrosine-protein kinase 7 (PTK7).

Application

Anti-PTK7, clone 31G9, Cat. No. MABS1923, is a rat monoclonal antibody that detects Inactive tyrosine-protein kinase 7 (PTK7) and has been tested for use in Flow Cytometry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunofluorescence Analysis: A 1:25 dilution from a representative lot detected PTK7 in human skin and human placenta tissues.

Immunofluorescence Analysis: A representative lot detected PTK7 in Immunofluorescence applications (Martinez, S., et. al. (2015). J Biol Chem. 290(51):30562-72).

Immunoprecipitation Analysis: A representative lot immunoprecipitated PTK7 in HEK293 cell lysate. (Martinez, S., et. al. (2015). J Biol Chem. 290(51):30562-72).

Western Blotting Analysis: A representative lot detected PTK7 in Western Blotting applications (Martinez, S., et. al. (2015). J Biol Chem. 290(51):30562-72).
Research Category
Signaling

Quality

Evaluated by Flow Cytometry in NCI-H460 cells.

Flow Cytometry Analysis: 1 µg of this antibody detected PTK7 in one million NCI-H460 cells.

Target description

118.40 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service