Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1823

Sigma-Aldrich

Anti-LATS1 Antibody, clone 9A3.1

clone 9A3.1, from mouse

Synonym(s):

Serine/threonine-protein kinase LATS1, EC: 2.7.11.1, Large tumor suppressor homolog 1, WARTS protein kinase, h-warts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9A3.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... LATS1(9113)

General description

Serine/threonine-protein kinase LATS1 (UniProt: O95835; also known as EC: 2.7.11.1, Large tumor suppressor homolog 1, WARTS protein kinase, h-warts) is encoded by the LATS1 (also known as WARTS) gene (Gene ID: 9113) in human. Serine/threonine-protein kinase LATS1 serves as a negative regulator of YAP1 in the Hippo signaling pathway, which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. YAP1 phosphorylation by LATS1 inhibits its translocation into the nucleus to regulate cellular genes that are important for cell proliferation, cell death, and cell migration. LATS1 is shown to be expressed in all adult tissues except in lung and kidney. LATS1 acts as a tumor suppressor and plays a critical role in maintenance of ploidy through its actions in both mitotic progression and the G1 tetraploidy checkpoint. It localizes to the centrosomes throughout interphase but migrates to the mitotic apparatus, including spindle pole bodies, mitotic spindle, and midbody, during mitosis. LATS1 negatively regulate G2/M transition by down-regulating CDK1 kinase activity. It is shown to be phosphorylated by STK3/MST2 at Serine 909 and Threonine 1079, which results in its activation. However, phosphorylation at Serine 464 by NUAK1 and NUAK2 leads to diminished protein level and it is essential for regulation of cellular senescence and cellular ploidy.

Specificity

Clone 9A3.1 specifically detects Serine/threonine protein kinase LATS1 in human cells. It targets an epitope within 116 amino acids from the N-termnal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 116 amino acids from the N-terminal half of human Serine/threonine-protein kinase LATS1.

Application

Anti-LATS1, clone 9A3.1, Cat. No. MABS1823, is a mouse monoclonal antiboody that detects Serine/threonine-protein kinase LATS1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected LATS1 in human prostate cancer and human cervical cancer tissue sections.

Western Blotting Analysis: 0.5 µg/mL from a representative lot detected LATS1 in K562 cell lysate.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected LATS1 in SH-SY5Y cell lysate.

Target description

~140 kDa observed; 126.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service