Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1814

Sigma-Aldrich

Anti-GATA2 Antibody, clone 3C10.1

clone 3C10.1, from mouse

Synonym(s):

Endothelial transcription factor GATA-2, GATA-binding protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
200 μL
$568.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
200 μL
$568.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C10.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GATA2(2624)

General description

Endothelial transcription factor GATA-2 (UniProt: P23769; also known as GATA-binding protein 2) is encoded by the GATA2 gene (Gene ID: 2624) in human. GATA2 is a zinc-finger transcriptional activator that regulates endothelin-1 gene expression in endothelial cells. It binds to the consensus sequence 5′-AGATAG-3′. GATA2 is expressed in embryonic brain and liver, endothelial cells, and in erythroid cells. GATA2 mutations appear to cause loss of function of the mutated allele leading to haploinsufficiency, which results in immunodeficiency characterized by significantly reduced or absent monocytes, B-lymphocytes, natural killer lymphocytes, and circulating and tissue dendritic cells. However, it does not affect the T-cell number. This deficiency leads to increased susceptibility to mycobacterial, papillomavirus, and opportunistic fungal infections. Certain mutations in GATA2 are also linked to lymphedema that results in swelling of the extremities due to altered lymphatic flow.

Specificity

Clone 3C10.1 specifically detects Endothelial transcription factor GATA-2 in human. It targets an epitope within 114 amino acids from the internal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 114 amino acids from the internal region of human GATA2.

Application

Anti-GATA2 Antibody, clone 3C10.1, Cat. No. MABS1814, is a mouse monoclonal antibody that targets GATA2 and has been tested in Western Blotting.

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GATA2 in 10 µg of SH-SY5Y cell lysate.

Target description

~52 kDa observed; 50.50 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service