Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1758

Sigma-Aldrich

Anti-RAD9A Antibody, clone 5H8.1

clone 5H8.1, from mouse

Synonym(s):

Cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, hRAD9, Rad9-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H8.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RAD9A(5883)

General description

Cell cycle checkpoint control protein RAD9A (EC 3.1.11.2; UniProt Q99638; also known as DNA repair exonuclease rad9 homolog A, hRAD9, Rad9-like protein) is encoded by the RAD9A (also known as RAD9A) gene (Gene ID 5883) in human. RAD9A is a component of the trimeric PCNA-like 9-1-1 complex (Rad9A, Rad1, and Hus1) and plays an important role in maintaining genomic integrity. RAD9A participates in both ATR and ATM signaling pathways-mediated cell cycle checkpoints in mitosis. Evidences indicate that RAD9A participates in DNA damage repair by interacting with proteins involved in base excision repair, mismatch repair and homologous recombination repair. RAD9A is shown to colocalize with γH2AX following DNA damage induction in an ATM-independent manner, and RAD9A inactivation delays the appearance of ionizing radiation-induced γH2AX foci. Rad9a-knockout causes embryonic lethality in mice due to increased apoptotic activity and reduced cellular proliferation, while mice heterozygous for Rad9a are prone to spontaneous and radiation-induced cataractogenesis. In addition, conditional knockout in keratinocytes causes enhanced susceptibility to carcinogen-induced skin tumors, while conditional Rad9a deletion in undifferentiated spermatogonia results in drastic effects on meiosis.

Specificity

Clone 5H8.1 targeted epitope lies within RAD9A ABL1-interacting domain.

Immunogen

GST-tagged recombinant human RAD9A ABL1-interacting domain fragment.

Application

Detect RAD9A using this mouse monoclonal Anti-RAD9A Antibody, clone 5H8.1, Cat. No. MABS1758, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected nuclear RAD9A immunoreactivity in human prostate cancer, pancreas, and small intestine tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected RAD9A in 10 µg of human skeletal muscle tissue lysate.

Target description

~42 kDa observed. 42.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse IgGκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service