Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1720

Sigma-Aldrich

Anti-Acetyl-CoA Synthetase Antibody, clone 6C1.2

clone 6C1.2, from mouse

Synonym(s):

Acetyl-coenzyme A synthetase, cytoplasmic, Acetate-CoA ligase, Acetyl-CoA synthetase, AceCS, ACS, Acyl-activating enzyme, Acyl-CoA synthetase short-chain family member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C1.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ACSS2(55902)

General description

Acetyl-coenzyme A synthetase, cytoplasmic (EC 6.2.1.1; UniProt Q9NR19; also known as Acetate-CoA ligase, Acetyl-CoA synthetase, AceCS, ACS, Acyl-activating enzyme, Acyl-CoA synthetase short-chain family member 2) is encoded by the ACSS2 (also known as ACAS2) gene (Gene ID 55902) in human. ACS plays a vital role in prokaryotic and eukaryotic cells by activating acetate to form acetyl CoA that serves as a source of two carbon metabolite for lipid synthesis and energy producing reactions. It is also reported to play a role in gene expression. In yeast cells it delivers acetyl CoA for histone acetylation. ACS contributes to cancer cell growth under low-oxygen and lipid-depleted conditions and its expression is upregulated under metabolically stress. Silencing ACSS2 gene is shown to reduce the growth of tumor xenografts. Ref.: Starai, VJ., and Escalante-Semerena, JC. (2004). Cell. Mol. Life Sci. 61, 2020 2030. Schug, ZT et al. (2015). Cancer Cell. 27:57-71.

Specificity

Immunogen sequence is present in both spliced isoforms of human acetyl-CoA synthetase reported by UniProt (Q9NR19).

Immunogen

GST-tagged recombinant human acetyl-CoA synthetase N-terminal fragment.

Application

Detect Acetyl-CoA synthetase using this mouse monoclonal Anti-Acetyl-CoA Synthetase Antibody, clone 6C1.2, Cat. No. MABS1720, validated for use in Western Blotting.

Quality

Evaluated by Western Blotting in HEPG2 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected acetyl-CoA synthetase in 10 µg of HepG2 cell lysate.

Target description

~80 kDa observed. 78.58/80.09 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service