Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1697

Sigma-Aldrich

Anti-WISP-3 Antibody, clone 13C4.1

clone 13C4.1, from mouse

Synonym(s):

WNT1-inducible-signaling pathway protein 3, CCN family member 6, WISP-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$570.00

$570.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$570.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$570.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13C4.1, monoclonal

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CCN6(8838)

General description

WNT1-inducible-signaling pathway protein 3 (UniProt O95389; also known as CCN family member 6, WISP-3) is encoded by the WISP3 (also known as CCN6, LIBC, PPAC, PPRD, UNQ462/PRO790/PRO956) gene (Gene ID 8838) in human. WISP-3 is a member of the CCN (CTGF, Cyr61, NOV) family connective tissue growth modulators involved in cell growth/differentiation during various phases of development through a complex interplay with BMP/Notch/Wnt signaling pathways. WISP-3 is expressed in cells and tissues of mesenchymal origin and WISP3 gene mutations are linked to an autosomal recessive skeletal disorder known as progressive pseudorheumatoid dysplasia (PPRD), indicating its importance in skeletal growth and the maintenance of cartilage integrity. WISP-3 interacts with BMP and components of the Wnt1 signaling pathway including LRP6 and Fz8, promoting the expression of cartilage-specific proteins collagen-II and aggrecan. Both IGF1 and ROS stimulate WISP-3 expression, WISP-3 in turn binds to and blocks IGF1 secretion, preventing IGF1-induced collagen X induction, ROS accumulation, and alkaline phosphatase activity, all of which are associated with chondrocyte hypertrophy induction. Human WISP-3 is synthesized with a signal peptide sequence (a.a. 1-23), the removal of which yields the mature protein containing several protein-interacting domains, including an IGF-interacting IGFBP domain (a.a. 44-117) with a von Willbrand factor type C repeat, a Thrombospondin (TSP) type-1 domain (a.a. 208-253), and a C-terminal cystine knot-like (CTCK) domain (a.a. 268-342).

Specificity

Clone 13C4.1 targets an epitope within the WISP-3 IGFBP domain.

Immunogen

GST-tagged recombinant human WISP-3 IGFBP domain fragment.

Application

Detect WISP-3 using this mouse monoclonal Anti-WISP-3, clone 13C4.1, Cat. No. MABS1697, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50-1,000 dilution of this antibody detected WISP-3 in human liver, kidney, and lung tissue sections.

Quality

Evaluated by Western Blotting in rat kidney tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected WISP-3 in 10 µg of rat kidney tissue lysate.

Target description

~39 kDa observed. 36.84/36.33 kDa (human/rat mature form), 39.29/41.40 kDa (human isoform 1/2 pro-form), and 38.92 kDa (rat pro-form) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service