Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1350

Sigma-Aldrich

Anti-Lipoprotein Lipase Antibody, clone 5D2

clone 5D2, from mouse

Synonym(s):

Lipoprotein lipase, LPL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5D2, monoclonal

species reactivity

rat, baboon, chicken, mink, feline, bovine, sheep, human, porcine, guinea pig

should not react with

mouse

technique(s)

ELISA: suitable
dot blot: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LPL(4023)
mouse ... Lpl(280843)

Related Categories

General description

Lipoprotein lipase (EC 3.1.1.34; UniProt P06858; also known as LPL) is encoded by the LPL gene (Gene ID 280843) in bovine species. Lipoprotein lipase (LPL) catalyzes the hydrolysis of triglycerides in plasma lipoproteins. LPL is produced by adipocytes and myocytes and secreted into the interstitial spaces, where it is bound by GPIHBP1 (a glycosylphosphatidylinositol-anchored protein of capillary endothelial cells) and shuttled to the luminal face of capillaries. The GPIHBP1 LPL complex is crucial for the binding of triglyceride-rich lipoproteins (TRLs) to endothelial cells and the subsequent lipolytic processing of TRLs. TRLs bind only the LPL-GPIHBP1 complex, but not GPIHBP1 alone, on the cell surface. A deficiency of either protein results in severe hypertriglyceridemia (chylomicronemia) and impaired delivery of lipid nutrients to parenchymal cells. Enzymatically active LPL appears to be a non-covalently linked homodimer with a head-to-tail subunit orientation that rapidly dissociates into inactive monomers. However, evidence for enzymatically active monomeric human LPL has also been presented. LPL is produced with a signal peptide sequence (a.a. 1-27), the removal of which yields the mature 448-amino acid (a.a. 28-475) enzyme containing a PLAT (Polycystin-1, Lipoxygenase, Alpha-Toxin) domain (a.a. 341-464) and a heparin-binding domain (a.a. 346-441).

Specificity

Clone 5D2 specifically reacts with LPL, but not the highly homologous hepatic lipase (HL) (Peterson, J., et al. (1992). J. Lipid Res. 33(8):1165-1170).

Immunogen

Purified bovine milk LPL.

Application

Detect Lipoprotein lipase using this mouse monoclonal Anti-Lipoprotein Lipase, clone 5D2 Antibody, Cat. No. MABS1350, validated for use in Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Inhibition assay, and Western Blotting.
Research Category
Signaling

Quality

Evaluated by Immunohistochemistry in human placenta tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected lipoprotein lipase/LPL in human placenta tissue sections.

Target description

50.55/53.38 kDa (bovine mature/proLPL) and 50.39/53.16 kDa (human mature/proLPL) calculated. ~56 kDa observed (Chang, S.F., et al. (1998). J. Lipid Res. 39(12):2350-2359; Peterson, J., et al. (1992). J. Lipid Res. 33(8):1165-1170).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shobini Jayaraman et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1867(1), 159064-159064 (2021-10-06)
Hydrolysis of VLDL triacylglycerol (TG) by lipoprotein lipase (LpL) is a major step in energy metabolism and VLDL-to-LDL maturation. Most functional LpL is anchored to the vascular endothelium, yet a small amount circulates on TG-rich lipoproteins. As circulating LpL has
Kohei Nishimoto et al.
Molecular metabolism, 40, 101025-101025 (2020-05-31)
Extrahepatic vitamin A is housed within organ-specific stellate cells that support local tissue function. These cells have been reported in the vocal fold mucosa (VFM) of the larynx; however, it is unknown how vitamin A reaches and is disseminated among

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service