Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS1280M

Sigma-Aldrich

Anti-mS2i6A Antibody, clone CL-6

clone CL-6, from mouse

Synonym(s):

N6-isopenetyl-2-thiomethyladenosine

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$599.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$599.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CL-6, monoclonal

species reactivity

mouse

species reactivity (predicted by homology)

mammals

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
dot blot: suitable

isotype

IgG2bκ

target post-translational modification

unmodified

General description

The mS2i6A is an evolutionally conserved modification found in the tRNA. The hypermodified mS2i6A at position 37 occurs frequently in tRNAs that read codons starting with uridine. The mS2 group of mS2i6A is critical for the accurate decoding of Tyr and Phe codons. It has been shown that a deficiency of mS2i6A in the cognate poly(U) reader tRNA(Phe) leads to increased misreading when the near-cognate competitor tRNA4(Leu) is wild-type. It can lead to higher rejection rate of tRNA4(Leu) by more aggressive proofreading on the wild-type ribosome. It is believed that Cdk5rap1 specifically catalyzes the mS2 modification of mt-tRNAs and contributes to mitochondrial function in vivo. This modification regulates efficient mitochondrial translation and energy metabolism in mammals. Cdk5rap1 knockout mice that are deficient in the 2-methylthio modification in mt-tRNAs bearing Ser(UCN), Phe, Tyr and Trp, impaired mitochondrial protein synthesis is observed, which causes respiratory defects of mitochondria and myopathy in vivo. (Ref.: Deuchler, M et al. (2016). Cell Mol Life Sci. 73 (16); 3075-3095; Fakruddin, M., et al. (2017). Nucleic Acids Res. 2017 Nov 16;45(20):11954-11961).

Specificity

Clone CL-6 recognizes wild-type total RNA, but not Cdk5rap1 KO total RNA.

Immunogen

A synthesized mS2i6A nucleotide conjugated to BSA.

Application

Anti-mS2i6A, clone CL-6, Cat. No. MABS1280, is a mouse monoclonal antibody that detects mS2i6A and has been tested for use in Dot Blot and ELISA.
Dot Blot Analysis: A 1:200 dilution of this antibody detected mS2i6A in Mouse liver total RNA and Cdk5rap1 KO mouse total RNA (Courtesy of Fan-Yan Wei, Ph.D., Faculty of Life Sciences, Kumamoto University Japan).

ELISA Analysis: A representative lot detected mS2i6A in ms2i6A and i6A wells (Courtesy of Fan-Yan Wei, Ph.D., Faculty of Life Sciences, Kumamoto University Japan).
Research Category
Signaling

Quality

Evaluated by Dot Blot in Mouse Liver Total RNA.

Dot Blot Analysis: A 1:2,000 dilution of this antibody detected mS2i6A in Mouse Liver total RNA.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service