Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1273

Sigma-Aldrich

Anti-Thrombomodulin Antibody, clone 411-201B

clone 411-201B, from rat

Synonym(s):

Thrombomodulin, CD141, Fetomodulin, SnoRNA MBII-339, TM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$523.00

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$523.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

411-201B, monoclonal

species reactivity

human, mouse

technique(s)

ELISA: suitable
electron microscopy: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... THBD(7056)

General description

Thrombomodulin (UniProt P15306; also known as CD141, Fetomodulin, SnoRNA MBII-339, TM) is encoded by the Thbd gene (Gene ID 21824) in murine species. Thrombomodulin (TM) is an endothelial cell surface protein that functions to suppress the propagation of thrombin (activated blood-coagulation factor II) generation by acting as a thrombin receptor. Formation of the TM-thrombin complex abolishes thrombin′s procoagulant activity by altering its substrate specificity, leading to proteolytic cleavage of protein C into the natural anticoagulant termed "activated protein C". Together with protein S, activated protein C proteolytically cleaves activated coagulation factors Va and VIIIa, thereby preventing the generation of activated coagulation factors Xa and thrombin. Murine thrombomodulin is initially produced with an N-terminal propeptide (a.a. 1-16) sequence, the removal of which yields the mature protein with a large extracellular region (a.a. 17-517), a transmembrane segment (a.a. 518-541), and a short cytoplamic tail (a.a. 542-577).

Immunogen

Epitope: Extracellular domain.
Normal mouse lung homogenate.

Application

Anti-Thrombomodulin Antibody, clone 411-201B is an antibody against Thrombomodulin for use in Immunohistochemistry (Paraffin), Electron Microscopy, Immunoprecipitation, Western Blotting, ELISA.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Thrombomodulin in human kidney tissue.
Electron Microscopy: A representative lot detected thrombomodulin immunoreactivity associated with capillary endothelial cells by EM using frozen mouse lung sections (Kennel, S.J., et al. (1988). Lab. Invest. 59(5):692-701).
Immunoprecipitation Analysis: A representative lot immunoprecipitated the 112 kDa thrombomodulin from normal mouse lung fibroblast (NMLF) lysates, but not from thrombomodulin-depleted NMLF lysates by performing IP with clone 273-34A (Cat. No. MABS1245) prior to IP with clone 411-201B (Kennel, S.J., et al. (1988). Lab. Invest. 59(5):692-701).
Western Blotting Analysis: A representative lot detected thrombomodulin (P112) in the normal mouse lung fibroblast (NMLF) immunoprecipitates obtained with either clone 411-201B or clone 273-34A (Cat. No. MABS1245) (Kennel, S.J., et al. (1988). Lab. Invest. 59(5):692-701).
Immunohistochemistry Analysis: Representative lots and clone 273-34A (Cat. No. MABS1245) showed similar staining pattern using both frozen and paraffin-embedded mouse lung sections (Ford, V.A., et al. (1992). J. Biol. Chem. 267(8):5446-5450; Rorvik, M.C., et al. (1988). J. Histochem. Cytochem. 36(7):741-749; Kennel, S.J., et al. (1988). Lab. Invest. 59(5):692-701).
ELISA Analysis: Representative lots were used as the detection antibody for the detection of thrombomodulin in mouse tissue samples by sandwich ELISA using clone 273-34A (Cat. No. MABS1245) as the capture antibody and an HRP-conjugated secondary antibody, or by sandwich radio-immunoassay employing 125I-labeled clone 411-201B without an enzyme-conjugated secondary antibody (Isermann, B., et al. (2001). Development. 128(6):827-838; Conway, E.M., et al. (1999). Blood. 93(10):3442-3450; Weiler-Guettler, H., et al. (1996). Circ. Res. 78(2):180-187; Ford, V.A., et al. (1992). J. Biol. Chem. 267(8):5446-5450).
Research Category
Signaling
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Immunohistochemistry in mouse lung tissue.

Immunohistochemistry Analysis: A 1:1,000 dilution of this antibody detected Thrombomodulin in mouse lung tissue.

Target description

61.87 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service