Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABS1235

Sigma-Aldrich

Anti-Progesterone Receptor A/B Antibody, clone 488/H3

clone 488/H3, from mouse

Synonym(s):

Progesterone receptor, PR, Nuclear receptor subfamily 3 group C member 3, Progesterone Receptor A/B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

488/H3, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PGR(5241)

General description

Progesterone receptor (UniProt P06401; also known as nuclear receptor subfamily 3 group C member 3, PR) is encoded by the PGR (also known as NR3C3) gene (Gene ID 5241) in human. The progesterone receptor (PR) is a hormone activated transcription factor, where hormone binding triggers the dissociation of heat shock and chaperone proteins from PR, leading to PR dimerization and activation. The receptor dimer then binds progestin response elements in the regulatory regions of its target genes and a multi-component complex is assembled to enable transcription. In addition, a number of kinases are reported to regulate PR activity via phosphorylation. PR can also regulate transcription without direct DNA binding by tethering to other transcription factors, such as specificity protein 1 (Sp1) or activator protein 1 (AP-1), and modulating their transcriptional activity on target genes. PR is expressed as five isoforms, including PR-B and PR-A that are produced as a result of alternate estrogen inducible promoters within the same PGR gene. PR-A lacks the first 164 amino acids of PR-B, but the two isoforms have otherwise identical sequence. PR-B and PR-A can regulate distinct subsets of target genes and studies in knockout mice suggest that PR-B predominantly regulates mammary gland development and PR-A is critical for normal uterine function.

Specificity

Clone 488/H3 reacts with both isoform 1 (isoform B; PR-B) and isoform 2 (isoform A; PR-A). This clone is also expected to react with isoform 5 (delta4), but not isoform 3 or 4 (PR-M).

Immunogen

Recombinant protein corresponding to the N-terminal half fragment of human Progesterone Receptor B.

Application

Immunocytochemistry Analysis: A representative lot detected exogenously expressed progesterone receptor isoform A and isoform B (PR-A & PR-B) in transfected MCF10A cells by fluorescent immunocytochemistry (Courtesy of Dr. Dean Edwards, Baylor College of Medicine, USA).
Western Blotting Analysis: A representative lot detected endogenous progesterone receptor isoforms A & B (PR-A & PR-B) in human breast cancer T47D cell lysate, as well as exogenously expressed PR-A & PR-B in transfected DCIS.COM human ductal carcinoma in situ (DCIS) breast cancer cells, but not in untransfected DCIS.COM cells (Courtesy of Dr. Dean Edwards, Baylor College of Medicine, USA).
This Anti-Progesterone Receptor A/B Antibody, clone 488/H3 is validated for use in Western Blotting and Immunocytochemistry for the detection of Progesterone Receptor A/B.

Quality

Evaluated by Western Blotting in human breast cancer T47D cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected both isoforms A and B (PR-A & PR-B) of progesterone receptor in human breast cancer T47D cell lysate.

Target description

~110/80 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service