Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1222

Sigma-Aldrich

Anti-MAN1B1 Antibody, clone 1D6

clone 1D6, from mouse

Synonym(s):

Endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase, MAN1B1, ER alpha-1,2-mannosidase, ER mannosidase 1, ERMan1, Man9GlcNAc2-specific-processing alpha-mannosidase, Mannosidase alpha class 1B member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D6, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MAN1B1(11253)

General description

Endoplasmic reticulum (ER) mannosyl-oligosaccharide 1,2-alpha-mannosidase (EC 3.2.1.113; UniProt Q9UKM7), also known as ER mannosyl-oligosaccharide 1,2-alpha-mannosidase, ER alpha-1,2-mannosidase, ERMan1, ERManI, ER mannosidase 1, Man9GlcNAc2-specific-processing alpha-mannosidase, Mannosidase alpha class 1B member 1) is encoded by the MAN1B1 gene in human. Originally thought to function in a similar fashion as its yeast ortholog Mns1p, which removes a terminal mannose unit to initiate a glycan-based ER-associated degradation (ERAD) signal. Mammalian MAN1B1 protein is actually localized in Golgi instead of ER. MAN1B1 protein participates in a Golgi-based quality control network that captures misfolded protein substrate, which escape from ER, most likely via COPII vesicles, and recycles them back to the ER for ERAD. Upon entering the Golgi, the initially escasped unfold proteins are retained by a cargo-capturing complex in which non-enzymatic regions of MAN1B1, including the decapeptide sequence (DPS) within the MAN1B1 luminal stem, play an integral role. Facilitated by direct interactions with gamma-COP, MAN1B1 promotes the recruitment of unfold proteins into COPI vesicles, which then recycles the unfold proteins back to the ER via retrograde COPI vesicles without the need for trimming by MAN1B1 protein.

Immunogen

GST-tagged recombinant protein corresponding to human MAN1B1.

Application

Anti-MAN1B1 Antibody, clone 1D6 is an antibody against MAN1B1 for use in Western Blotting, Immunoprecipitation, Immunocytochemistry.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected MAN1B1 in 10 µg of PC12 cell lysate.
Immunoprecipitation Analysis: A representative lot immunoprecipitated MAN1B1 in HeLa cell lysate ((Pan, S., et al. (2011). Mol. Bio. Cell. 22(16):2810-2822).
Immunoprecipitation Analysis: A representative lot immunoprecipitated MAN1B1 in transfected HeLa cells (Iannotti, M.J., et al. (2014). J. Biol. Chem. 289(17):11844-11858).
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected MAN1B1 in HeLa cells (Pan, S., et al. (2011). Mol. Bio. Cell. 22(16):2810-2822. & Iannotti, M.J., et al. (2014). J. Biol. Chem. 289(17):11844-11858).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected MAN1B1 in 10 µg of HeLa cell lysate.

Target description

~80/90 kDa observed. The calculated molecular weight of this protein is 80 kDa, but can be observed at ~90 kDa due to post translational modifications (Pan, S., et al. (2011). Mol. Bio. Cell. 22(16):2810-2822).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service