Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1193

Sigma-Aldrich

Anti-ABCB11 Antibody, clone 10D4.1

clone 10D4.1, from mouse

Synonym(s):

Bile salt export pump, ATP-binding cassette sub-family B member 11, Sister of P-glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10D4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ABCB11(8647)

General description

Bile salt export pump (UniProt O95342; also known as ATP-binding cassette sub-family B member 11, Sister of P-glycoprotein) is encoded by the ABCB11 (also known as BRIC2, BSEP, PFIC2, SPGP) gene (Gene ID 8647) in human. Bile is produced by hepatocytes in the liver and released into the biliary system of ducts and gallbladder. After food ingestion, bile is released in the form of mixed micelles of salts and phospholipids into the duodenum where its detergent properties act to solubilize lipids to help their absorption by the intestine. Bile salt export pump is an ATP-binding cassette (ABC) family member localized to the hepatocyte canalicular membrane where it plays an important role in the bile salt-dependent bile flow and the normal enterohepatic circulation of bile salts from the distal intestine back to the liver. ABCB11/BSEP gene mutations are associated with progressive familial intrahepatic cholestasis type 2 (PFIC2), benign recurrent intrahepatic cholestasis type 2 (BRIC2), drug-induced cholestasis, hormone-dependent intrahepatic cholestasis of pregnancy (ICP), biliary lithiasis, and transient neonatal cholestasis, as a result of impaired bile flow. Bile salt export pump is a 12-transmembrane protein (a.a. 63-83, 148-168, 216-236, 241-261, 320-340, 354-374, 756-776, 795-815, 870-890, 891-911, 980-1000, 1012-1032) with both its N- and C-terminal ends exposed cyroplasmically (a.a. 1-62 and 1033-1321).

Specificity

Clone 10D4.1 targets the fourth cytoplasmic domain of human ABCB11.

Immunogen

Epitope: The fourth cytoplasmic domain.
GST-tagged recombinant fragment derived from the fourth cytoplasmic domain of human ABCB11.

Application

Anti-ABCB11 Antibody, clone 10D4.1 is an antibody against ABCB11 for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected ABCB11 in human liver tissue sections.
Research Category
Signaling
Research Sub Category
Liver Physiology

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected ABCB11 in 10 µg of HepG2 cell lysate.

Target description

~150-175 kDa observed. 146.4 kDa calculated. The multiple banding pattern observed is consistent with the detection of the target protein with varying degrees of glycosylation.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service