Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN857

Sigma-Aldrich

Anti-SYNE1 Antibody, clone 200A6

clone 200A6, from mouse

Synonym(s):

CPG2, Enaptin, Enaptin-165, Nesprin-1 Giant, Syne-1B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

200A6, monoclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

GenBank accession no.

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Syne1(64009)

General description

Human SYNE1 gene (UniProt Q8NF91; Gene ID 23345) is among the top ten loci conferring risk for the five major psychiatric disorders, schizophrenia, bipolar disorder (BD), major depression, attention deficit-hyperactivity disorder (ADHD), and autism spectrum disorder (ASD), as well as multiple other neurological and neuromuscular disorders. The BD-associated single nucleotide polymorphisms (SNPs) are in a region of the human SYNE1 gene homologous to brain-specific transcripts of rat Syne1-encoding CPG2 (candidate plasticity gene 2; UniProt Q63128; Gene ID 499010), originally identified as an upregulated transcript during kainic acid-induced seizures in the rat dentate gyrus. The rat CPG2 spliced variant localizes to excitatory synapses of excitatory neurons, where it plays a role in synaptic plasticity and facilitates glutamate receptor cycling.

Specificity

Immunogen sequence is present in rat spliced isoform CPG2 (UniProt Q63128), as well as in mouse spliced isoform Enaptin (a.k.a. Nesprin-1 Giant), Syne-1B, and Enaptin-165 (UniProt Q6ZWR6). Target region is not found in rat isoform Nesprin-1 (UniProt Q8VHJ9), or in mouse isoform 2 and Syne-1A. Clone 200A6 detected shRNA-mediated knockdown of SYNE1 spliced isoform CPG2 in rat neurons by fluorescent immunocytochemistry and immunoblotting (Cottrell, J.R., et al. (2004). Neuron. 44(4):677-690).
This clone also detects SYNE1 protein in human brain samples with a much weaker signal than that observed in rat samples.

Immunogen

Synthetic peptide corresponding to a sequence within the third coiled coil of rat spliced isoform CPG2 or the tenth Spectrin domain of mouse spliced isoform 1 (a.k.a. Enaptin or Nesprin-1 Giant).

Application

Anti-SYNE1, clone 200A6, Cat. No. MABN857, is a highly specific mouse monoclonal antibody that targets SYNE1 splice variants, including rat CPG2, and has been tested in Electron Microscopy, Immunocytochemistry, and Western Blotting.
Electron Microscopy Analysis: A representative lot localized SYNE1 isoform CPG2 immunoreactivity concentrated in the postsynaptic site of excitatory synapses, in a distinct synaptic subdomain lateral to and beneath the postsynaptic density (PSD) by immunoEM labeling of 4% paraformaldehyde-/0.1% glutaraldehyde-fixed, lowicryl-embedded rat hippocampal cryosections (Cottrell, J.R., et al. (2004). Neuron. 44(4):677-690).

Electron Microscopy Analysis: A representative lot localized SYNE1 isoform CPG2 immunoreactivity colocalized with clathrin in the postsynaptic endocytic zone by immunoEM labeling of 60 nm thick serial sections of 4% paraformaldehyde-/0.1% glutaraldehyde-fixed rat neurons (Cottrell, J.R., et al. (2004). Neuron. 44(4):677-690).

Immunocytochemistry Analysis: A representative lot immunolocalized SYNE1 spliced isoform CPG2 on the dendritic spines, colocalized with clathrin, but segregated from the PSD and actin filaments in the spine head, by fluorescent immunocytochemistry staining of 4% formaldehyde-fixed, 0.3% Triton X-100-permeabilized rat hippocampal neurons. Target staining was greatly diminished among neurons received cellular CPG2 shRNA delivery (Cottrell, J.R., et al. (2004). Neuron. 44(4):677-690).

Western Blotting Analysis: A representative lot detected SYNE1 spliced isoform CPG2 as a ~110 kDa boublet band in rat cerebral cortex tissue extract and lysate from isolated rat cortical neurons. Target band detection was greatly diminished when using lysate from neurons following cellular CPG2 shRNA delivery (Cottrell, J.R., et al. (2004). Neuron. 44(4):677-690).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in rat hippocampus tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected the ~110 kDa SYNE1 spliced isoform CPG2 in 10 µg of rat hippocampus tissue lysate.

Target description

~110 kDa (rat isoform CPG2) observed. 109.0 kDa (rat isoform CPG2), 1,010 kDa (mouse isoforms Giant/Enaptin and Syne-1B), and 165.2 kDa (mouse isoform Enaptin-165) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service