Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN807

Sigma-Aldrich

Anti-EYS Antibody, clone 3G10.1

clone 3G10.1, from mouse

Synonym(s):

Protein eyes shut homolog, EGF-like protein 10, EGF-like protein 11, Epidermal growth factor-like protein 10, Epidermal growth factor-like protein 11, Protein spacemaker homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G10.1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EYS(346007)

General description

Protein eyes shut homolog (UniProt Q5T1H1; also known as EGF-like protein 10, EGF-like protein 11, Epidermal growth factor-like protein 10, Epidermal growth factor-like protein 11, Protein spacemaker homolog) is encoded by the EYS (also known as C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, SPAM) gene (ORF UNQ9424/PRO34591, Gene ID 346007) in human. Drosophila eyes shut protein is expressed in photoreceptor cells, the central nervous system, and peripheral sensory ganglia, and is essential for the formation of a matrix-filled intra-rhabdomeral space within the fly′s retina ommatidia, the loss of spacemaker (spam) converts an open rhabdom system to a closed one. Human EYS/SPAM gene likewise is found to be expressed in the eye and the spinal cord, and mutations in human EYS result in retinitis pigmentosa. Human EYS protein contains 27 EGF-like repeats and five laminin G-like (LG1-5) domains (between EGF repeats 21 & 21, 21 & 22, 23 & 24, 25 & 26, and after repeat EGF repeat 27). The presence of the EGF-like repeats is indicative of a role in the Notch signaling pathway, while EYS LG1-5 bear resemblance to the three agrin laminin G domains that are known to interact with several proteins, including α-dystroglycan at the muscle surface.

Specificity

Expected to react with spliced isoforms 1 and 3, but not isoforms 2 and 4, reported by UniProt (Q5T1H1).

Immunogen

Epitope: EGF-like domains 9-11.
GST-tagged recombinant human EYS fragment corresponding to the EGF-like domains 9-11.

Application

Anti-EYS Antibody, clone 3G10.1 is an antibody against EYS for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected EYS in human and rat retina tissue sections.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Quality

Evaluated by Western Blotting of EYS recombinant protein.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected EYS recombinant protein.

Target description

348.4 kDa (isoform 1) and 350.8 kDa (isoform 3) kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service