Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN793

Sigma-Aldrich

Anti-PTHrP (1-34) Antibody, clone 23-57-137-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$513.00

$513.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$513.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$513.00


Please contact Customer Service for Availability

Request a Bulk Order

antibody form

purified antibody

Quality Level

clone

#23-57-137-1
monoclonal

purified by

using protein G

species reactivity

mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

western blot: suitable

isotype

IgG1κ

capacity

neutralization

UniProt accession no.

General description

Bovine thyroglobulin-conjugated linear peptide corresponding to human PTHrP(1-34).

Specificity

This monoclonal detects PTHrP(1-34)/PTHrP(1-36), but not PTHrP(38-94) or Osteostatin.

Application

Epitope: PTHrP(1-34)

Quality

Evaluated by Western Blotting in human PTHrP recombinant protein.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PTHrP(1-34) in 1 µg of human PTHrP recombinant protein.

Physical properties

~10 kDa observed

Physical form

Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service