Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN791

Sigma-Aldrich

Anti-VAP-B Antibody, clone 3C6.2

clone 3C6.2, from mouse

Synonym(s):

Vesicle-associated membrane protein-associated protein B, VAMP-associated protein B, VAMP-B, VAP-B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$591.00

$591.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$591.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$591.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3C6.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... VAPB(9217)

General description

Vesicle-associated membrane protein-associated protein B (UniProt O95292; also known as VAMP-associated protein B, VAMP-B, VAP-B) is encoded by the VAPB (also known as ALS8, UNQ484/PRO983) gene (Gene ID 9217) in human. VAMP-associated proteins (VAPs) are type IV membrane proteins that are well conserved among species. There exist three human VAPs encoded by two genes, with VAPA encoding VAP-A and VAPB encoding VAP-B and VAP-C. VAPs generally localize at the endoplasmic reticulum (ER), although they are also reported to localize at other subcellular organelles in some species and cell types. VAPs were shown to have important roles in non-vesicular lipid transport, lipid metabolism, the regulation of ER structure, and the unfolded protein response through MSP domain-mediated interaction with FFAT motifs. VAP-B functions as an adaptor protein to recruit target proteins to the ER and execute various cellular functions, including lipid transport, membrane traffic, and ER stress response. VAP-B binds the catalytic subunit Rab3 GTPase activating protein 1 (Rab3GAP1), but not the non-catalytic subunit Rab3GAP2 of the heterodimeric Rab3GAP complex. VAP-B and Rab3GAP interaction is mediated through the Rab3GAP1 acidic tract (FFAT)-like motif and is essential for VAP-B-dependent regulation of the nuclear envelope protein transport through the ER-Golgi intermediate compartment (ERGIC). VAP-B contains a major sperm protein (MSP; a.a. 7-124) domain at the N-terminal cytoplasmic region, followed by a coiled-coil region (a.a. 2-222), and a transmembrane (TM) domain (a.a. 223-243).

Specificity

Clone 3C6.2 epitope lies within VAP-B extracellular coiled coil region not present in isoform VAP-C.

Immunogen

GST-tagged recombinant fragment from the extracellular coiled coil region of human VAP-B.

Application

Anti-VAP-B, clone 3C6.2, Cat. No. MABN791, is a highly specific mouse monoclonal antibody that targets VAMP-associated protein B (VAP-B) and has been tested in Immunostochemistry (Paraffin) and Western Blotting applications.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected VAP-B in human brain and kidney tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected VAP-B in 10 µg of HeLa cell lysate.

Target description

~29 kDa observed. 27.10 kDa calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service