Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN787

Sigma-Aldrich

Anti-Dual oxidase 2, clone Duox S-12 Antibody

clone Duox S-12, from mouse

Synonym(s):

Dual oxidase 2, Large NOX 2, Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, Thyroid oxidase 2, p138 thyroid oxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Duox S-12, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DUOX2(50506)

General description

Dual oxidase 2 is an important enzyme in thyroid hormone biosynthesis and peroxide metabolism. Dual oxidase 2 generates the hydrogen peroxide needed for thyroid peroxidase and lactoperoxidase enzymes as well as processes H2O2 made during metabolic processes. Dual oxidase also plays a role in anti-microbial defense in our mucous membranes by providing the peroxidase for lactoperoxidase’s antimicrobial activities. Dual oxidase 2 is also needed for various extracellular matrix stabilizations and protein cross-linking needs such as during cuticle formation. Dual oxidase 2 is widely expressed in the developing fetus, and in the adult, it is largely expressed in the mucosal and epithelial cells and linings of the gut, lung, salivary glands and testis. Dual oxidase also plays a role in many hypersensitivity and inflammatory diseases and is upregulated in diseases such as colitis and certain cancers.

Immunogen

Epitope: N-terminus
Recombinant protein corresponding to the N-terminus of human Dual oxidase 2.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Dual oxidase 2 in human prostate cancer and human breast cancer tissue.
Western Blotting Analysis: A representative lot detected Dual oxidase 2 in BxPC-3 cell lysate (Wu, Y., et al. (2013). International Journal of Oncology. 42:1229-1238).
Immunocytochemistry Analysis: A representative lot detected Dual oxidase 2 in MIA PACA-2 cell (Wu, Y., et al. (2013). International Journal of Oncology. 42:1229-1238).
Immunohistochemistry Analysis: A representative lot detected Dual oxidase 2 MIA PACA-2 tissue (Wu, Y., et al. (2013). International Journal of Oncology. 42:1229-1238).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
This Anti-Dual oxidase 2, clone Duox S-12 Antibody is validated for use in Western Blotting and Immunohistochemistry and Immunocytochemistry for the detection of Dual oxidase 2.

Quality

Evaluated by Western Blotting in BxPC3 treated with IL-4 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Dual oxidase 2 in 10 µg of BxPC3 treated with IL-4 cell lysate.

Target description

~175 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darrin V Bann et al.
Cancer research, 79(21), 5490-5499 (2019-09-11)
Highly penetrant hereditary thyroid cancer manifests as familial nonmedullary thyroid cancer (FNMTC), whereas low-penetrance hereditary thyroid cancer manifests as sporadic disease and is associated with common polymorphisms, including rs965513[A]. Whole-exome sequencing of an FNMTC kindred identified a novel Y1203H germline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service