Skip to Content
MilliporeSigma
All Photos(3)

Documents

MABN740

Sigma-Aldrich

Anti-GABA-B receptor 2 Antibody, clone N81/37

clone N81/37, from mouse

Synonym(s):

GABA-B-R2, GABA-BR2, GABABR2, Gamma-aminobutyric acid type B receptor subunit 2, GABA-B receptor 2, Gb2, G-protein coupled receptor 51

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

N81/37, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GABBR2(9568)

General description

Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the mammalian central nervous system. GABA exerts its effects through ionotropic [GABA(A/C)] receptors, to produce fast synaptic inhibition, and metabotropic [GABA(B)] receptors, to produce slow, prolonged inhibitory signals. The GABA(B) receptor consists of a heterodimer of two related 7-transmembrane receptors, GABA(B) receptor 1 and GABA(B) receptor 2. The GABA(B) receptor 1 gene is mapped to chromosome 6p21.3 within the HLA class I region close to the HLA-F gene. Susceptibility loci for multiple sclerosis, epilepsy, and schizophrenia have also been mapped in this region. Alternative splicing of this gene generates 4 transcript variants.

Specificity

This antibody does not cross react with GABABR1 (Prof. J. Trimmer, University of California, Davis).

Immunogen

Recombinant protein corresponding to rat GABA-B receptor 2.

Application

Anti-GABA-B receptor 2 Antibody, clone N81/37 is a highly specific mouse monoclonal antibody, that targets GABA B Receptor & has been tested in western blotting & IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected GABA-B receptor 2 in 10 µg of mouse brain tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected GABA-B receptor 2 in human cerebellum and rat cerebral cortex tissue.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected GABA-B receptor 2 in 10 µg of rat brain tissue lysate.

Target description

~105 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service