Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN696

Sigma-Aldrich

Anti-G6PD Antibody, clone 2H7

ascites fluid, clone 2H7, from mouse

Synonym(s):

Glucose-6-phosphate dehydrogenase, G6PD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$500.00

$500.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$500.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$500.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2H7, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... G6PD(2539)

Related Categories

General description

Glucose-6-Phosphate Dehydrogenase (G6PD) is the rate-limiting enzyme in the pentose phosphate pathway that catalyzes the oxidation of catalyzes the oxidation of glucose-6-phosphate (G6P) to form pentose sugars for subequent biosynthetic pathways. Glucose-6-Phosphate Dehydrogenase also produces nicotinamide adenine dinucleotide phosphate (NADPH), an important electron donor that plays a protective role against oxidative stress, especially in red blood cells. Deficiencies in glucose-6-phosphate dehydrogenase may disrupt cell growth, cell signaling, and embryonic development. It is also fairly well established that mutations in the glucose-6-phosphate dehydrogenase result in a highly varied condition known as chronic non-spherocytic hemolytic anemia.

Immunogen

Purified recombinant fragment of human G6PD expressed in E. Coli.

Application

Detect Glucose-6-Phosphate Dehydrogenase using this mouse monoclonal antibody, Anti-G6PD Antibody, clone 2H7 validated for use in western blotting, IHC & Flow Cytometry.
Immunohistochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected G6PD in human breast cancer and kidney cancer tissues.

ELISA (Direct) Analysis: A 1:10,000 dilution from a representative lot detected G6PD with recombinant G6PD protein antigen.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected G6PD in non serum starved Jurkat cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in HeLa, MCF-7, Jurkat and K562 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected G6PD in HeLa, MCF-7, Jurkat and K562 cell lysates.

Target description

~59 kDa observed. Uncharacterized bands may appear in some lysate(s).

Linkage

Replaces: MABS391

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa, MCF-7, Jurkat and K562 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service