Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN645

Sigma-Aldrich

Anti-CD39 Antibody, clone 5E12

clone 5E12, 1 mg/mL, from rat

Synonym(s):

Ectonucleoside triphosphate diphosphohydrolase 1, NTPDase 1, Ecto-ATP diphosphohydrolase 1, Ecto-ATPDase 1, Ecto-ATPase 1, Ecto-apyrase, Lymphoid cell activation antigen, CD39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5E12, monoclonal

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Entpd1(12495)

Related Categories

General description

CD39 (EC 3.6.1.5; also known as Ecto-apyrase, Ecto-ATPase 1, Ecto-ATPDase1 , Ecto-ATP diphosphohydrolase 1, Ectonucleoside triphosphate diphosphohydrolase 1, Lymphoid cell activation antigen, NTPDase 1) is encoded by the Entpd1 or Cd39 gene (Entrez Gene ID 12495) in murine species. CD39 is a cell surface nucleotide-metabolizing enzyme that hydrolyzes extracellular purine NTP and NDP. E. coli infection is reported to induce CD39 promoter activity in macrophages, resulting in an upregulated CD39 expression during sepsis in mice, while CD39 knockdown increases mortality among mice with polymicrobial sepsis (PMID 25318479). Anti-CD39, clone 5E12 binds the murine CD39 (P55772) extracellular domain and is demonstrated to selectively stain murine macroglia by flow cytometry (PMID 22863620).

Immunogen

Epitope: extracellular domain
Whole cells with surface CD39.

Application

Anti-CD39 Antibody, clone 5E12 is an antibody against CD39 for use in Flow Cytometry.
Flow Cytometry Analysis: A representative lot detected CD39 in CD11b+ cells (Butovsky, O., et al. (2012). J. Clin. Invest. 122(9):3063-3087).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Flow Cytometry in CD11b+ mouse splenocytes.

Flow Cytometry Analysis: Various dilutions of this antibody detected CD39 in CD11b+ mouse splenocytes.

Target description

70-100 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service