MABN633
Anti-α-Synuclein (SNCA) Antibody
mouse monoclonal, 10D2
Synonym(s):
Alpha-synuclein, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP
Select a Size
$534.00
Select a Size
About This Item
$534.00
Recommended Products
Product Name
Anti-a-Synuclein, clone 10D2 Antibody, clone 10D2, from mouse, purified by affinity chromatography
biological source
mouse
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
10D2, monoclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
ELISA: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SNCA(6622)
General description
This antibody can be used as a sandwich ELISA pair with Cat. No. MABN389, Anti-Aggregated a-Synuclein, clone 5G4.
Immunogen
Application
ELISA Analysis: A representative lot from an independent laboratory detected aggregated a-Synuclein in a sandwich ELISA with Cat. No. MABN389, Anti-Aggregated a-Synuclein, clone 5G4 (Kovacs, G. G., et al. (2012). Acta Neuropathol. 123(1):37-50.).
Quality
Western Blotting Analysis: 1.5 µg/mL of this antibody detected a-Synuclein in 10 µg of human brain tissue lysate.
Target description
Linkage
Other Notes
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service