Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN615

Sigma-Aldrich

Anti-Pan-GRK Antibody, clone N145/20

clone N145/20, from mouse

Synonym(s):

G protein-coupled receptor kinase 5, G protein-coupled receptor kinase GRK5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

N145/20, monoclonal

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Grk5(14773)

General description

G protein-coupled receptor kinases (GRK) are serine/threonine kinases that phosphorylate a variety of G-protein-coupled receptors (GPCRs), specifically activated forms, including adrenergic receptors, muscarinic acetylcholine receptors, dopamine receptors and opioid receptors. GRK receptor phosphorylation initiates beta-arrestin-mediated receptor desensitization, internalization, and signaling events leading to their down-regulation.

Specificity

This antibody recognizes the C-terminus of GRK. This antibody cross reacts with GRK6 (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to the C-terminus of mouse Pan-GRK.

Application

Anti-Pan-GRK Antibody, clone N145/20 Cat. No. MABN615 is a highly specific mouse monoclonal antibody & that targets GRK & has been tested in western blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse liver tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Pan-GRK in 10 µg of mouse liver tissue lysate.

Target description

~ 68 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service