Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN540

Sigma-Aldrich

Anti-mGluR5 Antibody, clone N75/33

clone N75/33, from mouse

Synonym(s):

Metabotropic glutamate receptor 5, mGluR5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$564.00

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N75/33, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRM5(2915)

General description

Metabotropic glutamate receptor 5 (mGluR5) is a G-protein mediated glutamate receptor that generates a calcium-activated chloride current. mGluR5 is widely distributed in neuronal cells of the central nervous system.

Specificity

This antibody recognizes the C-terminus of mGluR5. This antibody may cross react with mGluR1 based upon <35% sequence homology.

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to the C-terminus of rat mGluR5.

Application

Detect mGluR5 using this mouse monoclonal antibody, Anti-mGluR5 Antibody, clone N75/33 validated for use in western blotting & IHC.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western Blotting Analysis: 1 µg/mL from a representative lot detected mGluR5 in 10 µg of rat brain membrane tissue.

Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected mGluR5 in normal human midbrain tissue.

Quality

Evaluated by Western Blotting in mouse hypothalamus tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected mGluR5 in 10 µg of mouse hypothalamus tissue lysate.

Target description

~150-270 kDa observed. This protein may be observed at higher molecular weights due to glycosylation (Bhave, G., et al. (2003). J Biol Chem. 278(32):30294-30301). This protein is observed as a dimer at ~270 kDa, but is observed as a monomer at ~140 kDa under reduced conditions in SDS-PAGE (Romano, C., et al. (1996). J Biol Chem. 271(45):28612-28616.). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Hypothalamus tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cameron W Morris et al.
Biological psychiatry, 93(11), 976-988 (2023-02-24)
Grooming dysfunction is a hallmark of the obsessive-compulsive spectrum disorder trichotillomania. Numerous preclinical studies have utilized SAPAP3-deficient mice for understanding the neurobiology of repetitive grooming, suggesting that excessive grooming is caused by increased metabotropic glutamate receptor 5 (mGluR5) activity in
Wilber Romero-Fernandez et al.
Molecular neurobiology, 59(10), 5955-5969 (2022-07-14)
The adenosine A2A receptor (A2AR), dopamine D2 receptor (D2R) and metabotropic glutamate receptor type 5 (mGluR5) form A2AR-D2R-mGluR5 heteroreceptor complexes in living cells and in rat striatal neurons. In the current study, we present experimental data supporting the view that
Ming Chen et al.
Molecular neurodegeneration, 16(1), 14-14 (2021-03-06)
Before the deposition of amyloid-beta plaques and the onset of learning memory deficits, patients with Alzheimer's disease (AD) experience olfactory dysfunction, typified by a reduced ability to detect, discriminate, and identify odors. Rodent models of AD, such as the Tg2576
Caesar M Hernandez et al.
eNeuro, 5(3) (2018-07-05)
Glutamate signaling is essential for the persistent neural activity in prefrontal cortex (PFC) that enables working memory. Metabotropic glutamate receptors (mGluRs) are a diverse class of proteins that modulate excitatory neurotransmission via both presynaptic regulation of extracellular glutamate levels and
Qingwei Huo et al.
Cerebral cortex (New York, N.Y. : 1991), 27(8), 4060-4072 (2016-08-25)
Genetic and biochemical studies have focused on the role of amyloid β protein in the pathogenesis of Alzheimer's disease. In comparison, the physiological roles of its precursor protein, amyloid precursor protein (APP), in synaptic and network activity is less well studied.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service