Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN349

Sigma-Aldrich

Anti-Synaptotagmin-12 Antibody (SYT12), clone 1B12.1

clone 1B12.1, 0.5 mg/mL, from mouse

Synonym(s):

Synaptotagmin-12, Synaptotagmin XII, SytXII, Synaptotagmin-12 (SYT12)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$399.00

$399.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$399.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$399.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B12.1, monoclonal

species reactivity

human, rat

concentration

0.5 mg/mL

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SYT12(91683)

General description

Synaptotagmin-12 (SYT12) , also known as Synaptotagmin-12, Synaptotagmin XII, SytXII, and encoded by the gene SYT12, is an important protein involved in calcium dependent exocytosis of secretory vesicles through calcium and phospholipid binding to the C2 domain and Synaptotagmin-12 may serve as calcium sensors in the process of vesicular trafficking and exocytosis. Synaptotagmin-12 is a synaptic vesicle phosophoprotein that modulates spontaneous neurotransmitter release, but unlike other family members Synaptotagmin-12 differs from classical synaptotagmins in that it does not bind Ca(2+). In synaptic vesicles, synaptotagmin-12 forms a complex with synaptotagmin-1 that prevents synaptotagmin-1 from interacting with SNARE complexes. Synaptotagmin-12 also is an abundant synaptic vesicle protein that is important in LTP transmission. For instance, its phosphorylation by cAMP Protein Kinase is necessary for hippocampal mossy fiber LTP.

Immunogen

Epitope: Cytoplasmic domain
GST-tagged recombinant protein corresponding to the cytoplasmic domain of human Synaptotagmin-12 (SYT12).

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Synaptotagmin-12 Antibody (SYT12), clone 1B12.1 is validated for use in Western Blotting for the detection of Synaptotagmin-12 (SYT12).

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Synaptotagmin-12 (SYT12) in 200 µg of rat brain tissue lysate.

Target description

~ 47 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service