Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN2678

Sigma-Aldrich

Anti-phospho-α-synuclein (Ser129) Antibody, clone PS129

new

Synonym(s):

Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor (NACP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

PS129, monoclonal

mol wt

calculated mol wt 14.46 kDa
observed mol wt ~16 kDa

purified by

using protein G

species reactivity

human, mouse

species reactivity (predicted by homology)

bovine, porcine, rat

packaging

antibody small pack of 100

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... SNCA(6622)

Specificity

Clone PS129 is a mouse monoclonal antibody that specifically detects -synuclein phosphorylated on serine 129.

Immunogen

KLH-conjugated linear peptide corresponding to 9 amino acids surrounding phosphoserine 129 from the C-terminal region of human -synuclein.

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant -synuclein phosphorylated S129 and recombinant -synuclein monomers.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected -synuclein phosphorylated on serine 129 in recombinant -synuclein phosphorylated S129 protein, but not in recombinant -synuclein monomers.

Tested Applications

ELISA Analysis: A representative lot detected -synuclein in ELISA application (Majbour, N.K., et al. (2016). Mol Neurodegener. 11:7; El-Agnaf, O., et al. (2017). Neurobiol Dis. 104: 85-96).

Immunohistochemistry Application: A representative lot detected phospho- -synuclein (Ser129) in Immunohistochemistry application (Vaikath, N.N., et al. (2015). Neurobiol Dis. 79:81-99).

Western Blotting Analysis: A representative lot detected phospho- -synuclein (Ser129) in Western Blotting application (Majbour, N.K., et al. (2016). Mol Neurodegener. 11:7).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Alpha-synuclein (UniProt: P37840; also known as Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, NACP) is encoded by the SNCA (also known as NACP, PARK1) gene (Gene ID: 6622) in human. Alpha-synuclein is a membrane-bound protein in dopaminergic neurons that is involved in the regulation of dopamine release and transport. It is also reported to induce fibrillization of microtubule-associated protein tau and reduces neuronal responsiveness to various apoptotic stimuli. It is primarily expressed in the brain and is associated with the regulation of neurotransmitter release at synapses. Alpha-synuclein is also found in the cytoplasm, where it exists in a dynamic equilibrium between a soluble cytosolic form and a membrane-bound form. It associates with lipid membranes, particularly those of synaptic vesicles. Aggregation of alpha-synuclein is known to be a causative factor in the development of Parkinson′s disease (PD), which leads to the degeneration of dopaminergic neurons in the substantia nigra (SN) in the midbrain region. -synuclein can be phosphorylated, predominantly on serine residues by casein kinase 1 (CK1). Phosphorylation by CK1 appears to occur on residue (serine 129) distinct from the residue (tyrosine 125) phosphorylated by other kinase FYN. The phosphorylation of -synuclein at serine 129 (pS129) has been extensively studied due to its significant implications in neurodegenerative diseases, particularly Parkinson′s disease (PD) and other synucleinopathies. Phosphorylation of Ser129 is reported to be selective and extensive in synucleinopathy lesions and promotes insoluble fibril formation. This serves as a potential biomarker for disease diagnosis and progression and offers a promising target for therapeutic intervention. (Ref.: Vaikath, N.N., et al. (2015). Neurobiol Dis. 79:81-99; Majbour, N.K., et al. (2016). Mol Neurodegener. 19:11:7; El-Agnaf, O., et al. (2017). Neurobiol Dis. 104:85-96).

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service