Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN2613

Sigma-Aldrich

Anti-RTN/NOGO-A Antibody, clone NG1

Synonym(s):

Anti-RTN/NOGO-A Antibody, clone NG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

hamster

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

NG1, monoclonal

mol wt

calculated mol wt 126.61 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Rtn4(68585)

General description

Reticulon-4 (UniProt: Q99P72; also known as Neurite outgrowth inhibitor, Nogo protein) is encoded by the Rtn4 (also known as Kiaa0886, Nogo) gene (Gene ID: 68585) in murine species. NOGO proteins are required for the formation and stabilization of endoplasmic reticulum (ER) tubules. They are also involved in nuclear envelope expansion, nuclear pore complex formation and proper localization of inner nuclear membrane proteins. Three isoforms of NOGO have been described that share a common C-terminal domain but differ in their N-terminal sequence. NOGO-A is the longest isoform and is considered as a developmental neurite growth factor. It acts as a negative regulator of axon-axon adhesion and growth, but also acts as a facilitator of neurite branching. In the adult central nervous system (CNS), it is involved in down-regulation of growth, stabilization of wiring, and restriction of plasticity. It can also serve as a negative regulator of CNS angiogenesis by blocking spreading, migration, and sprouting of primary brain microvascular endothelial cells. Isoform B mainly functions in the endothelial cells and vascular smooth muscle cells. It promotes the migration of endothelial cells but inhibits the migration of vascular smooth muscle cells. Isoform C regulates cardiomyocyte apoptosis under hypoxic conditions. Clone NG1 recognizes NOGA-A but does not react with other isoforms. It is shown to strongly bind olfactory bulb axons in the lateral olfactory tract as well as some other growing axons in nervous system. This clone has been use to show that knockdown of NOGO-A reduces the number of axonal collateral branches in olfactory bulb. (Ref.: Iketani, M., et al. (2016). Sci. Rep. 6; 39586; Petrinovic, MM., et al. (2010). Development. 137(15); 2539-2550; Tozaki, H., et al. (2002). Mol. Brain Res. 104(2); 111-119).

Specificity

Clone NG1 is an Armenian Hamster monoclonal antibody that detects murine Reticulon-4 (RTN4).

Immunogen

Lateral olfactory tract of E14.5 mouse embryos fixed with 4% paraformaldehyde in phosphate-buffered saline (PBS), washed and suspended in PBS.

Application

Quality Control Testing

Evaluated by Immunohistochemistry in E14 mouse embryo frozen tissue sections.

Immunohistochemistry Applications: A 1:200 dilution of this antibody detected RTN/NOGO-A in E14 mouse embryo frozen tissue sections.

Tested Applications

Immunohistochemistry Application: A representative lot detected RTN/NOGO-A in Immunohistochemistry applications (Iketani, M., et al. (2016). Sci Rep. 6:39586; Tozaki, H., et al. (2002). Brain Res Mol Brain Res. 104(2):111-9).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-RTN/NOGO-A, clone NG1, Cat. No. MABN2613, is an Armenian hamster monoclonal antibody that detects Reticulon-4 and is tested for use in Immunohistochemistry.

Physical form

Purified armenian hamster monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service