Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2490

Sigma-Aldrich

Anti-mutant HTT Antibody, clone PHP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

PHP1
monoclonal

purified by

using protein G

species reactivity

mouse, human, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

dot blot: suitable
electron microscopy: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

Mode of action

| inhibits

General description

GST-tagged recombinant exon1 of wild-type human huntingtin HTTx1 (20Q) protein.

Specificity

Clone PHP1 is mouse monoclonal antibody that detects mutated Huntingtin protein. It targets an epitope QAQPLLPQP within the proline-rich domain.

Application

Western Blotting Analysis: A representative lot detected mutant HTT in Western Blotting applications (Ko, J., et. al. (2018). Hum Mol Genet. 27(13):2330-2343).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected mutant HTT in brain tissue lysates of 6 and 9 month-old WT or Q140 KI HD mice (Courtesy of Ali Khoshnan, Ph.D., California Institute of Technology, Paadena, CA).

Electron Microscopy Analysis: A representative lot detected mutant HTT in Immuno-Electron Microscopy applications (Ko, J., et. al. (2018). Hum. Mol. Genet. 27(13): 2330-2343).

Inhibits Activity/Function Analysis: A representative lot blocked seeding and subsequent fibril assembly of mHTTx1 in vitro and in a cell culture model of HD. (Ko, J., et. al. (2018). Hum Mol Genet. 27(13):2330-2343).

Immunohistochemistry Analysis: A representative lot detected mutant HTT in Immunohistochemistry applications (Ko, J., et. al. (2018). Hum Mol Genet. 27(13): 2330-2343).

Dot Blot Analysis: A representative lot detected mutant HTT in Dot Blot applications (Ko, J., et. al. (2018). Hum Mol Genet. 27(13):2330-2343).

Quality

Evaluated by Immunohistochemistry (Paraffin) in Human Huntington′s diseased brain tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected mutant HTT in Human Huntington′s diseased brain tissue sections.

Physical properties

347.60 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Human, Mouse.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service