Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2487

Sigma-Aldrich

Anti-Astrin Antibody, clone C-1

clone C-1, from mouse

Synonym(s):

Sperm-associated antigen 5, Deepest, Mitotic spindle-associated protein p126, MAP126

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C-1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... SPAG5(10615)

General description

Sperm-associated antigen 5 (UniProt: Q96R06; also known as Astrin, Deepest, Mitotic spindle-associated protein p126, MAP126) is encoded by the SPAG5 gene (Gene ID: 10615) in human. Astrin is a homodimeric protein with a globular head domain and a long stalk. It is an essential component of the mitotic spindle. Astrin is highly expressed in testis and is detected at low levels in placenta, liver, pancreas, thymus and colon. Its expression is cell cycle-regulated, with an increase from prophase to cytokinesis and return to basal levels at the next G1 phase. In non-dividing cells Astrin is associated with microtubules unless under cellular stress, then it is localized in stress granules. In non-mitotic cells, upon stress induction, it inhibits mTORC1 association and recruits the mTORC1 component RPTOR to stress granules, thereby preventing mTORC1 hyperactivation-induced apoptosis. Astrin is required for normal chromosome segregation and progression into anaphase. It is also required for chromosome alignment, normal timing of sister chromatid segregation, and maintenance of spindle pole architecture. In complex with SKAP, it promotes stable microtubule-kinetochore attachments. Depletion of astrin in cells is shown to result in the activation of p53-dependent apoptosis that elevates Bax expression and increases Caspase-3 activity. Astrin can be phosphorylated at multiple sites by Aurora kinase A. (Ref.: Yang, YC., et al. (2006). Biochem. Biophys. Res. Commun. 343(2); 428-434).

Specificity

Clone C-1 specifically detects human Sperm-associated antigen 5 (Astrin). It detects both splice variants (130 and 140 kDa).

Immunogen

His-tagged recombinant fragment corresponding to the first 952 amino acids from human Sperm-associated antigen 5 (Astrin).

Application

Anti-Astrin, clone C-1, Cat. No. MABN2487, is a highly specific mouse monoclonal antibody that targets Sperm-associated antigen 5 and has been tested for use in Immunocytochemistry and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Astrin in Western Blotting applications (Yang, Y.C., et. al. (2006). Biochem Biophys Res Commun. 343(2):428-34).

Immunocytochemistry Analysis: A representative lot detected Astrin in Immunocytochemistry applications (Schmidt, J.C., et. al. (2010). J Cell Biol. 191(2):269-80; Yang, Y.C., et. al. (2006). Biochem Biophys Res Commun. 343(2):428-34; Kern, D.M., et. al. (2016). J Cell Biol. 213(3):315-28).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Astrin in HeLa cell lysate.

Target description

~130 and 140 kDa doublet; 134.42 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrik Risteski et al.
Cell reports, 40(5), 111169-111169 (2022-08-05)
Chromosome alignment at the spindle equator promotes proper chromosome segregation and depends on pulling forces exerted at kinetochore fiber tips together with polar ejection forces. However, kinetochore fibers are also subjected to forces driving their poleward flux. Here we introduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service