Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2436

Sigma-Aldrich

Anti-Guanylate Cyclase 2E Antibody, clone 8A5

clone 8A5, from mouse

Synonym(s):

RETGC-1, Guanylate cyclase-E, Guanylate cyclase-2E, GC-E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8A5, monoclonal

species reactivity

bovine, mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Gucy2E(14919)

General description

Guanylyl cyclase GC-E (UniProt: P52785; also known as EC: 4.6.1.2, Guanylate cyclase 2E, GC-2E) is encoded by the Gucy2e (also known as Guc2e) gene (Gene ID: 14919) in murine species. GC-2E is a single-pass type I membrane protein that plays a specific functional role in the rods and/or cones of photoreceptors. It may be the enzyme involved in the re-synthesis of cGMP required for recovery of the dark state after phototransduction. Following photoexcitation, cGMP in photoreceptors is first hydrolyzed by a light-activated phosphodiesterase (PDE6) and then replenished by a retinal membrane guanylyl cyclase. It is synthesized with a 50 amino acid signal peptide that is cleaved to generate the mature enzyme. It consists of an extracellular domain (aa 51-469), a helical domain (aa 470-490) and a cytoplasmic domain (aa 491-1108).

Specificity

Clone 8A5 detects Guanylate cyclase-1 in Bovine, Human, and murine species. It targets an epitope within 48 amino acids from the C-terminal end.

Immunogen

Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 48 amino acids from the C-terminal end of human Guanylate cyclase-1.

Application

Anti-Guanylate Cyclase-2E, clone 8A5, Cat. No. MABN2436, is a highly specific mouse monoclonal antibody that targets retinal Guanylyl cyclase (RETGC-1; GC-2E) and has been used in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Guanylate Cyclase-1 in mouse retina tissues (Courtesy of Dr. Robert Molday, Ph.D., University of British Columbia, Canada).

Immunofluorescence Analysis: A representative lot detected Guanylate Cyclase-1 in Immunofluorescence applications (Molday, L.L., et. al. (2013). Hum Mol Genet. 22(19):3894-905).

Western Blotting Analysis: 10 ug/mL of a representative lot detected Guanylate Cyclase-1 in mouse retina tissue lysates (Courtesy of Dr. Robert Molday, Ph.D., University of British Columbia, Canada).

Western Blotting Analysis: A representative lot detected Guanylate Cyclase-1 in Western Blotting applications (Peshenko, I.V., et. al. (2011). Biochemistry. 50(44):9511-9; Molday, L.L., et. al. (2013). Hum Mol Genet. 22(19):3894-905)
Research Category
Neuroscience

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Target description

~120 kDa observed; 120.06 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service