Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN2435

Sigma-Aldrich

Anti-Peripherin-2 Antibody, clone 5H2

clone 5H2, from mouse

Synonym(s):

Retinal degeneration slow protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H2, monoclonal

species reactivity

bovine, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Prph2(19133)

Related Categories

General description

Peripherin-2 (UniProt: P17810; also known as Retinal degeneration slow protein) is encoded by the PRPH2 (also known as RDS) gene in bovine species. Peripherin-2 is a disulfide-linked homodimeric, multi-pass membrane protein that may function as an adhesion molecule involved in stabilization and compaction of outer segment disks in retina and is considered to be essential for disk morphogenesis. It is exclusively expressed in outer segments. Peripherin-2 can form homo- and heteromeric protein complexes in outer segment. The core homomeric peripherin-2 unit is the non-covalent tetramer, which can also assemble into covalently linked octamers and higher-order oligomers. The homomeric peripherin-2 complexes are essential for proper outer segment morphology and architecture. Peripherin-2 is also shown to form heteromeric complexes with its homolog ROM-1. Mutations in PRPH2 gene can lead to autosomal dominant retinitis pigmentosa (adRP) And a majority of these mutations are located within the large loop domain connecting the transmembrane domain 3 and 4 (Ref.: Bohm, S et al. (2017). Scientific Reports 7, Article number: 2321).

Specificity

Clone 5H2 detects Peripherin-2 in bovine and mouse retina.

Immunogen

Epitope: C-terminus
Purified bovine whole peripherin.

Application

Anti-Peripherin-2, clone 5H2, Cat. No. MABN2435, is a highly specific mouse monoclonal antibody that targets Peripherin-2 and has been tested in Immunofluorescence, Radioimmunoassay, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Peripherin-2 in Western Blotting applications (Connell, G., et. al. (1991). Proc Natl Acad Sci USA. 88(3):723-6; Zhang, Y., et. al. (2009). J Cell Sci. 122(Pt 8):1192-200).

Radioimmunoassay Analysis: A representative lot detected Peripherin-2 in Radioimmunoassay applications (Connell, G., et. al. (1991). Proc Natl Acad Sci USA. 88(3):723-6).

Immunofluorescence Analysis: A representative lot detected Peripherin-2 in Immunoflourescence applications (Zhang, Y., et. al. (2009). J Cell Sci. 122(Pt 8):1192-200).

Quality

Evaluated by Western Blotting in bovine retinal tissue lysate.

Western Blotting Analysis: 4 µg/mL of this antibody detected Peripherin-2 in 10 µg of bovine retinal tissue lysate.

Target description

~39 kDa observed; 38.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service