Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN2433

Sigma-Aldrich

Anti-TMEM16B Antibody, clone 13B8

clone 13B8, from mouse

Synonym(s):

Anoctamin-2, Transmembrane protein 16B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13B8, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Ano2(243634)

Related Categories

General description

Anoctamin-2 (UniProt: Q8CFW1; also known as Transmembrane protein 16B, TMEM16B) is encoded by the Ano2 (also known as Tmem16b) gene (Gene ID: 243634) in murine species. TMEM16B is a multi-pass membrane protein that serves as a calcium-activated chloride channel (CaCC) that plays a role in olfactory signal transduction and also mediates light perception amplification in retina. Its channel activity is shown to be repressed by chloride channel inhibitors, such as niflumic acid and flufenaic acid. TMEM16 B is expressed in retina, especially in the photoreceptor synaptic terminals, and in olfactory epithelium, particularly in sensory neurons (OSNs) and cilia. It is also shown to be expressed in retinal pigment epithelium (RPE), olfactory bulb, and cortex. It is detected in the mantle layer of the neural tube and in the dorsal root ganglia at E14.5. TMEM16B is also involved in a calcium-dependent mode of ionic plasticity that reduces the efficacy of GABAergic synapses. It is reported to attenuate GABAergic transmission by increasing the postsynaptic chloride concentration, hence reducing the driving force for chloride influx.

Specificity

Clone 13B8 detects Transmembrane protein 16B (TMEM16B) in murine neural tissue.

Immunogen

GST-tagged recombinant fragment corresponding to the first 74 amino acids of murine Transmembrane protein 16B.

Application

Anti-TMEM16B, clone 13B8, Cat. No. MABN2433, is a highly specific mouse monoclonal antibody that targets Transmembrane protein 16B (Anoctamin-2) and has been tested in Immunofluorescence and Western Blotting.
Western Blotting Analysis: A representative lot detected TMEM16B in mouse retina tissue (Courtesy of Dr. Robert Molday, Ph.D., University of British Columbia, Canada).

Immunofluorescence Analysis: A representative lot detected TMEM16B in mouse retina tissue (Courtesy of Dr. Robert Molday, Ph.D., University of British Columbia, Canada).

Quality

Evaluated by Immunohistochemistry in mouse retinal tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected TMEM16B in mouse retinal tissue.

Target description

130 kDa observed; 114.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service