Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN2416M

Sigma-Aldrich

Anti-HSD17B10 Antibody, clone 5F3

clone 5F3, from mouse

Synonym(s):

3-hydroxyacyl-CoA dehydrogenase type-2, 17-beta-hydroxysteroid dehydrogenase 10, 17-beta-HSD 10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, 3-hydroxyacyl-CoA dehydrogenase type II, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein,

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$231.00
100 μG
$546.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$231.00
100 μG
$546.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F3, monoclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... HSD17B10(3028)

General description

3-hydroxyacyl-CoA dehydrogenase type-2 (UniProt: Q99714; also known as 17-beta-hydroxysteroid dehydrogenase 10, 17-beta-HSD 10, 3-hydroxy-2-methylbutyryl-CoA dehydrogenase, 3-hydroxyacyl-CoA dehydrogenase type II, Endoplasmic reticulum-associated amyloid beta-peptide-binding protein, Mitochondrial ribonuclease P protein 2, Mitochondrial RNase P protein 2, Short chain dehydrogenase/ reductase family 5C member 1, Short-chain type dehydrogenase/reductase XH98G2, Type II HADH) is encoded by the HSD17B10 (also known as ERAB, HADH2, MRPP2, SCHAD, SDR5C1, XH98G2) gene (Gene ID: 3028) in human. 3-hydroxyacyl-CoA dehydrogenase type-2 is a part of mitochondrial ribonuclease P, an enzyme composed of MRPP1/TRMT10C, MRPP2/HSD17B10 and MRPP3/KIAA0391, which cleaves tRNA molecules in their 5′-ends. It functions in mitochondrial tRNA maturation. It catalyzes the beta-oxidation at position 17 of androgens and estrogens and has 3-alpha-hydroxysteroid dehydrogenase activity with androsterone. It catalyzes the third step in the beta-oxidation of fatty acids and also carries out oxidative conversions of 7-alpha-OH and 7-beta-OH bile acids. With C21 steroids it exhibits 20-beta-OH and 21-OH dehydrogenase activities. May contribute to neuronal dysfunction associated with Alzheimer disease (AD) via interactions with intracellular amyloid-beta peptides. 3-hydroxyacyl-CoA dehydrogenase type-2 is ubiquitously expressed in normal tissues, but is overexpressed in neurons affected in AD.

Specificity

Clone 5F3 detects 3-hydroxyacyl-CoA dehydrogenase type-2 in human and murine cells.

Immunogen

GST-tagged full length recombinant human 3-hydroxyacyl-CoA dehydrogenase type-2 protein (ERAB/HADH).

Application

Anti-HSD17B10 antibody, clone 5F3, Cat. No. MABN2416, is a highly specific mouse monoclonal antibody that targets HSD17B10 and has been tested for use in Electron Microscopy, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Electron Microscopy Analysis: A representative lot detected HSD17B10 in Electron Microscopy applications (Wen, G.Y., et. al. (2002). Brain Res. 954(1):115-22).

Immunocytochemistry Analysis: A representative lot detected HSD17B10 in Immunocytochemistry applications (Frackowiak, J., et. al. (2001). Brain Res. 907(1-2):44-53).

Western Blotting Analysis: A representative lot detected HSD17B10 in Western Blotting applications (Frackowiak, J., et. al. (2001). Brain Res. 907(1-2):44-53).

Immunohistochemistry Analysis: A 1:250-1:1,000 dilution from a representative lot detected HSD17B10 in human placenta and human breast cancer tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293T cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected HSD17B10 in 10 µg of HEK293T cell lysate.

Target description

~27 kDa observed; 26.92 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service