Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2398

Sigma-Aldrich

Anti-Klotho Antibody, clone KL-115

clone KL-115, from rat

Synonym(s):

EC: 3.2.1.31, KL

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$231.00
100 μL
$629.00

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$231.00
100 μL
$629.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$231.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

antibody form

unpurified

antibody product type

primary antibodies

clone

KL-115, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... KL(9365)

General description

Klotho (UniProt: Q9UEF7; also known as EC: 3.2.1.31) is encoded by the KL gene (Gene ID:9365) in human. Klotho is a homodimeric, highly N-glycosylated, single-pass type I membrane protein that is present in kidney, placenta, small intestine, and prostate tissue. In kidney it is predominantly present in cortical renal tubules. A soluble secreted form (isoform 2, ~ 64 kDa) is present in serum and cerebrospinal fluid. The Klotho peptide generated by cleavage of the membrane-bound isoform (Isoform 1) is considered as an anti-aging circulating hormone that is reported to extend life span by inhibiting insulin/IGF1 signaling. It also acts as a cofactor for interaction of FGF23 with FGF R1, which negatively regulates the activity of 1-alpha-hydroxylase, the rate limiting enzyme in the synthesis of 1,25(OH)2D3. Klotho is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to generate the mature form that has an extracellular domain (aa 31-981), a short transmembrane domain (aa 982-1002), and a short cytoplasmic tail (aa 1003-1012). Deficiency of Klotho is reported to shorten life-span in animal models with a myriad of disorders associated with old age, such as arteriosclerosis, osteoporosis, and skin atrophy. Klotho deficient mice also display severe hyperphosphatemia and soft tissue calcification. On the contrary, over-expression of Klotho is shown to extend life-span in murine models by about 30% with side effects, such as mild insulin resistance and enhanced resistance to oxidative stress. Defects in KL gene has been linked to chronic renal failure complications.

Specificity

Clone KL-115 is a rat monoclonal antibody that specifically detects human Klotho. It targets an epitope with in 207 amino acids from the KL1 domain from the N-terminal region.

Immunogen

GST-tagged recombinant fragment corresponding to 207 amino acids from the extracellular domain from the N-terminal region of human Klotho.

Application

Anti-Klotho, clone KL-115, Cat. No. MABN2398, is a highly specific rat monoclonal antibody that targets klotho and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A represetative lot detected Klotho in Western Blotting applications (Maltare, A., et. al. (2014). Monoclon Antib Immunodiagn Immunoother. 33(6):420-7).

Immunocytochemistry Analysis: A represetative lot detected Klotho in Immunocytochemistry applications (Maltare, A., et. al. (2014). Monoclon Antib Immunodiagn Immunoother. 33(6):420-7).

Immunoprecipitation Analysis: A represetative lot immunoprecipitated Klotho in Immunoprecipitation applications (Maltare, A., et. al. (2014). Monoclon Antib Immunodiagn Immunoother. 33(6):420-7).

Quality

Evaluated by Western Blotting in human kidney tissue lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Klotho in 10 µg of human kidney tissue lysates.

Target description

~130 kDa observed; 116.18 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rat monoclonal antibody in ascites with 0.1% sodium azide with 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service