MABN2311
Anti-NUMB Antibody, clone Ab21
Synonym(s):
Protein numb homolog;h-Numb;Protein S171
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
Ab21, monoclonal
mol wt
calculated mol wt 70.8 kDa
observed mol wt ~70 kDa
purified by
using protein G
species reactivity
human, mouse
species reactivity (predicted by homology)
monkey
packaging
antibody small pack of 100 μL
technique(s)
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
C-terminal half
Protein ID accession no.
UniProt accession no.
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... NUMB(8650)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Neuro2a cell lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected NUMB in Neuro2a cell lysate.
Tested Applications
Western Blotting Analysis: A 1:500 dilution from a representative lot detected NUMB in A549 and U87MG cell lysates.
Immunofluorescence Analysis: A representative lot detected NUMB in Immunofluorescence application (Colaluca, I.N., et al. (2008). Nature. 451(7174):76-80; Tosoni, D., et al. (2015). J Cell Biol. 211(4):845-62; Santoro, A., et al. (2019). Cell Rep. 26(3):624-638.e8).
Immunoprecipitation Analysis: A representative lot immunoprecipitated NUMB in Immunoprecipitation application (Colaluca, I.N., et al. (2008). Nature. 451(7174):76-80; Colaluca, I.N., et al. (2018). J Cell Biol. 217(2):745-762).
Western Blotting Analysis: A representative lot detected NUMB in Western Blotting application (Colaluca, I.N., et al. (2008). Nature. 451(7174):76-80; Tosoni, D., et al. (2015). J Cell Biol. 211(4):845-62; Tosoni, D., et al. (2017). EMBO Mol Med. 9(5):65-671; Colaluca, I.N., et al. (2018). J Cell Biol. 217(2):745-762).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service