Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN2304

Sigma-Aldrich

Anti-Legumain/Asparaginyl endopeptidase antibody Clone 6E3

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$239.00
100 μG
$603.00

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$239.00
100 μG
$603.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

$239.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6E3, monoclonal

mol wt

calculated mol wt 49.41 kDa
observed mol wt ~55 kDa

purified by

using protein G

species reactivity

human, mouse

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... LGMN(5641)

General description

Legumain (UniProt: Q99538; also known as EC: 3.4.22.34, Asparaginyl endopeptidase, AEP, Protease cysteine 1, Delta Secretase) is encoded by the LGMN (also known as PRSC1) gene (Gene ID: 5641) in human. AEP is a lysosomal endopeptidase that displays strict specificity for the hydrolysis of asparaginyl bonds and can simultaneously cleave both amyloid precursor protein (APP) and tau. It cleaves aspartyl bonds slowly, especially under acidic conditions. AEP is synthesized with a signal peptide (aa 1-17) and a propeptide (aa 324-433) that are cleaved off in the mature form. It exists a homodimer before removal of the propeptide and after autocatalytic processing exists as a monomer. In its zymogen form, the uncleaved propeptide blocks access to the active site. AEP is phosphorylated on serine 226 by serine-arginine protein kinase 2 (SRPK2), which accelerates its autocatalytic cleavage and enhances its enzymatic activity. The active form translocates to cytoplasm. In Alzheimer s disease brains AEP is reported to be highly phosphorylated. Depletion of AEP is shown to reduce amyloid peptide production and senile plaque formation in 5XFAD mouse brain that leads to restoration of synaptic activity and cognitive function. Three isoforms of AEP have been described that are produced by alternative splicing. (Ref.: Wang Z-H., et al. (2017). Mol. Cell 67(5); 812-825; Zhang, Z et al (2015). Nat. Commun. 6; 9762).

Specificity

Clone 6E3 is a mouse monoclonal antibody that specifically detects Legumain (AEP).

Immunogen

His-tagged full-length recombinant human legumain.

Application

Quality Control Testing

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected Legumain/AEP in SH-SY5Y cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Legumain/AEP in mouse brain tissue lysate.

ELISA Analysis: A representative lot detected Legumain/AEP in ELISA applications (Mattock, K.L., et al. (2010). Atherosclerosis.208(1):83-9; Kang, S.S., et al. (2018). EMBO J. 37(12):e98878).

Immunoprecipitation Analysis: A representative lot immunoprecipitated legumain/AEP in Immunoprecipitation applications (Zhang, Z., et al. (2015). Nat Commun. 6:8762).

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Legumain/AEP in human placenta tissue sections.

Immunocytochemistry Analysis: A representative lot detected Legumain/AEP in Immunocytochemistry applications (Li, D.N., et al. (2003). J. Biol. Chem. 278(40):38980-90; Wang, Z-H., et al. (2019). Cell Rep. 28(3):655-669).

Western Blotting Analysis: A representative lot detected Legumain/AEP in Western Blotting applications (Probst-Kepper, M., et al. (2009). J Cell Mol Med.;13(9B):3343-57; Zhang, Z., et al. (2015). Nat Commun.;6:8762; Wang, Z-H., et al. (2017). Mol Cell.;67(5):812-825; Zhang, Z., et al. (2017). Nat Struct Mol Biol.;24(8):632-642; Kang, S.S., et al. (2018). EMBO J.;37(12):e98878; Wang, Z-H., et al. (2018). Nat Commun.;9(1):1784; Wang, Z-H., et al. (2018). JCI Insight.;3(16):e99007; Wang, Z-H., et al. (2019). Cell Rep.;28(3):655-669).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-LGMN/AEP, clone 6E3, Cat. No. MABN2304, is a mouse monoclonal antibody that detects Legumain and is tested for use in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service