Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABN2277

Sigma-Aldrich

Anti-Dab1 Antibody, clone 4H11

clone 4H11, from rat

Synonym(s):

Disabled homolog 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H11, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Dab1(13131)

Related Categories

General description

Disabled homolog 1 (UniProt: P97318; also known as DAB1) is encoded by the Dab1 gene (Gene ID: 13131) in murine species. Dab 1 is mainly expressed in the brain and serves as an adapter molecule involved in neural development where it is reported to direct the migration of cortical neurons past previously formed neurons to reach their proper layer. It is a cytosolic protein that is shown to be essential component of the reelin signaling pathway. It is shown to bind to the cytoplasmic tail of Reelin receptor, APO E receptor 2, and to LDL receptor. Dab1 can be phosphorylated on Tyr198, Tyr220, and Tyr232 upon reelin induction in embryonic neurons. It can also be phosphorylated on Ser524, but this phosphorylation is independent of reelin signaling. Tyrosine phosphorylation of Dab1 by Src family tyrosine kinases is considered to be the most critical downstream event in Reelin signaling. Dab1 contains a phosphotyrosine interaction domain (PID) (aa 36-189), which specifically binds to the Asn-Pro-Xaa-Tyr(P) motif found in many tyrosine-phosphorylated proteins. Seven different isoforms of Dab1 have been identified that are generated via alternative splicing. Clone 4H1 binds to an epitope within amino acids 281-460. (Ref.: Onoue, A., et al. (2014). Neurosci. Res. 88; 23-27).

Specificity

Clone 4H11 is a rat monoclonal antibody that detects murine Disabled homolog 1 (Dab1).

Immunogen

GST-tagged full-length recombinant murine Disabled homolog 1 expressed in E. coli.

Application

Anti-Dab1, clone 4H11, Cat. No. MABN2277, is a highly specific rat monoclonal antibody that targets Disabled homolog 1 and has been tested for use in Immunoprecipitation, Immunohistochemistry, and Western Blotting.
Immunohistochemistry Analysis: A representative lot detected Dab1 in Immunohistochemistry applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-7).

Western Blotting Analysis: A representative lot detected Dab1 in Western Blotting applications (Onoune, A., et. al. (2014). Neurosci Res. 88:23-7; Nakamura, K., et. al. (2016). Neuroscience. 336:20-29).

Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Dab1 in cerebral cortex tissue lysate of P20 mouse (Courtesy of Prof. Mitsuharu Hattori, Nagoya City University).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293T cells expressing murine Dab 1.


Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Dab1 in lysates from HEK293T cells expressing murine Dab 1 but not in mock HEK293T cells.

Target description

68 kDa observed; 63.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service