Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN1790

Sigma-Aldrich

Anti-C9ORF72/C9RANT (poly-AP) Antibody, clone 14E2

clone 14E2, from rat

Synonym(s):

C9ORF72/C9RANT (poly-PA)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$523.00

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$523.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$523.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14E2, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Expansion of a GGGGCC hexanucleotide repeat sequence in the non-coding region of human chromosome 9 open reading frame 72 or C9orf72 (also known as ALSFTD, FTDALS; Gene ID 203228) is the most common genetic abnormality in familial and sporadic frontotemporal dementia (FTD) and motor neuron disease (MND), with amyotrophic lateral sclerosis (ALS) as the most frequent form. The number of hexanucleotide repeats in the normal population ranges from 2 to 24, whereas up to several thousand repeats (700 - 4,400 repeats) are found in the pathologically expanded allele. Unconventional repeat-associated non-ATG-initiated translation (RANT) of the GGGGCC repeats in the three alternate reading frames from both sense and antisense transcripts generates five types of dipeptide repeat (DPR) protein species, each composed of repeating units of two amino acids, glycine-alanine (GA), glycine-proline (GP), glycine-arginine (GR), alanine-proline (AP), proline-arginine (PR), respectively. Immunofluorescence localization of exogenously epxressed DPR-GFP fusions in HEK293 transfectants showed dot-like or star-shaped poly-GA inclusions with predominant cytoplasmic and occasional nuclear localization, while poly-GR and poly-PR showed large dot-like intranuclear inclusions and occasional smaller cytoplasmic inclusions. In contrast, poly-PA was evenly distributed throughout the nucleus and cytoplasm without apparent aggregation.

Specificity

Clone 14E2 recognizes C9ORF72/C9RANT (poly-AP) and other proteins containing poly-AP (poly-PA) sequence.
Target sequence is not species-specific.

Immunogen

Epitope: poly-AP (poly-PA).
Recombinant GST fusion with a 10 AP-repeat sequence (GST-AP10).

Application

Anti-C9ORF72 Antibody, clone 14E2 is an antibody against C9RANT (poly-AP) for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected the expression an exogenously transfected PA175-myc dipeptide repeat (DPR) construct containing a 175 PA-repeat sequence by fluorescent immunocytochemistry staining of 4% paraformaldehyde/4% sucrose-fixed, 0.3% Triton X-100-permeablized HEK293 transfectants (May, S., et al. (2014). Acta Neuropathol. 128(4):485-503).
Western Blotting Analysis: A representative lot detected the expression of an exogenously transfected PA175-myc dipeptide repeat (DPR) construct containing a 175 PA-repeat sequence in lysate from HEK293 transfectants (May, S., et al. (2014). Acta Neuropathol. 128(4):485-503).

Quality

Evaluated by Western Blotting iof recombinant GST fusion with a 10 AP-repeat sequence (GST-AP10).

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 3 µg of recombinant GST fusion with a 10 AP-repeat sequence (GST-AP10).

Target description

Variable. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service