Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABN1623

Sigma-Aldrich

Anti-FGF-9 Antibody, clone 3A10.1

clone 3A10.1, from mouse

Synonym(s):

Fibroblast growth factor 9, FGF-9, GAF, Heparin-binding growth factor 9, Glia-activating factor, HBGF-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A10.1, monoclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... FGF9(2254)

General description

Fibroblast growth factor 9 (UniProt P31371; also known as FGF-9, GAF, Heparin-binding growth factor 9, Glia-activating factor, HBGF-9) is encoded by the FGF9 (also known as SYNS3) gene (Gene ID 2254) in human. Fibroblast growth factor 9 (FGF9) belongs to the FGF family of proteins that regulate a variety of cell functions, including proliferation, differentiation, survival, and motility, via FGF receptors (FGFRs). FGF-9 interacts with several FGFRs and binds preferentially to FGFR3. FGF-9 is highly expressed in cancer-associated fibroblasts (CAFs) in gastric cancer and may play a pivotal role in the interaction between cancer cells and their surrounding stromal cells. FGF-9 is shown to induce MMP-7 expression in AGS and MKN28 gastric cancer cells, promoting invasion and cellular anti-apoptotic activity through activation of the Erk and Akt signaling pathways. In human brain, FGF-9 is primarily expressed by neurons in the cortex, hippocampus, thalamus, cerebellum, and spinal cord, as well as by glia in the hindbrain and spinal cord. Upregulated FGF-9 expression is found in postmortem hippocampus tissues from major depressive disorder (MDD) patients, while FGF-2 is downregulated in the depressed brain. Likewise, chronic intracerebroventricular administration of FGF-9 increased both anxiety- and depression-like behaviors in rats, while FGF-9 knockdown in the dentate gyrus of the hippocampus ameliorated anxiety-like behavior. Human FGF-9 is produced with a propeptide (a.a. 1-3), which is cleaved off posttranslationally to yield the mature protein.

Specificity

Clone 3A10.1 targets an internal sequence conserved among bovine, human, mouse, and rat species.

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the C-terminal half of human FGF-9.

Application

Anti-FGF-9, clone 3A10.1 , Cat. No. MABN1623, is a highly specific mouse monoclonal antibody that targets FGF-9 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected FGF-9 in human cerebellum, cerebral cortex, and pancreas tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting of recombinant human FGF-9.

Western Blotting Analysis: 2 µg/mL of this antibody detected 50 ng of E. coli expressed full-length recombinant human FGF-9.

Target description

~23 kDa (non-glycosylated recombinant mature hFGF-9) observed. 23.14/23.44 kDa (human mature/pro-form), 23.08/23.38 kDa (bovine mature/pro-form), 23.11/23.41 kDa (mouse & rat mature/pro-form), 23.16/23.46 kDa (porcine mature/pro-form) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service