Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN118

Sigma-Aldrich

Anti-Limk1 Antibody, clone 6C3.1| MABN118

clone 6C3.1, from mouse

Synonym(s):

LIM domain kinase 1, LIMK-1, Lim kinase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$564.00

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$564.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$564.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C3.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIMK1(3984)

General description

LIM Kinase 1 (LIMK1) is a protein kinase that plays an important role in cellular functions like cell motility and growth cone actin dynamics. LIMK1 contains one PDZ domain and two LIM domains which mediate protein to protein inteactions. It is widely expressed in the adult and fetal nervous system with the highest levels being found in the cerebral cortex. Mutations in LIMK1 have been associated with cardiovascular and musculo-skeletal abnormalities.

Immunogen

GST-tagged recombinant protein corresponding to human Limk1.

Application

Detect Limk1 using this Anti-Limk1 Antibody, clone 6C3.1 validated for use in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Limk1 in human thalamus and human cerebral cortex tissue.

Quality

Evaluated by Western Blotting in human cortex tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Limk1 in 10 µg of human cortex tissue lysate.

Target description

~72 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service