Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF958

Sigma-Aldrich

Anti-CD47 Antibody, clone PF3.1

clone PF3.1, from mouse

Synonym(s):

Leukocyte surface antigen CD47, Antigenic surface determinant protein OA3, CD47, IAP, Integrin-associated protein, Protein MER6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$629.00

$629.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$629.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$629.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PF3.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CD47(961)

Related Categories

General description

Leukocyte surface antigen CD47 (UniProt Q08722; also known as Antigenic surface determinant protein OA3, CD47, IAP, Integrin-associated protein, Protein MER6) is encoded by the CD47 (also known as MER6) gene (Gene ID 861) in human. Originally defined as a tumor-specific marker for ovarian carcinoma, CD47 a multitransmembrane glycoprotein widely expressed on all hematopoietic cells and most other cell types. CD47 plays a role in αvβ3-mediated cell adhesion and Ca2+ influx, as well as in platelet activation via interaction with thrombospondin-1. In addtion, CD47 modulates peripheral mononuclear (PMN) neutrophil migration across endothelial and epithelial monolayers. The rate of fMLP-stimulated PMN migration is shown to be decreased by CD47 functional blockage upon neutralizaing antibody treatment, which can be partially reversed by PI3K inhibition. Following fMLP stimulation and after PMN transmigration, secondary granules-derived CD47 molecules are shown to accumulate on plasma membrane, resulting in increased cell surface CD47 immunoreactivity. CD47 is produced with a signal peptide sequence (a.a. 1-18) that is removed posttranslationally to yiled the 5-transmembrane (a.a. 142-162, 177-197, 208-228, 236-256, 269-289) protein with its N-terminal end (a.a. 19-141), composed almost entirely of a V-type Ig-like domain (a.a. 19-127), exposed extracellularly and its C-terminal end (a.a. 290-323) at the cytoplasmic side.

Specificity

Clone PF3.1 detects human CD47 by targeting CD47 extracellular domain. In contrast to clone C5/D5 (Cat. No. MABF959), clone PF3.1 is suitable for immunoblotting, but not suitable for neutralizing CD47-dependent cellular signaling, while both clones can stain cell surface CD47 (Liu, Y., et al. (2001). J. Biol. Chem. 276(43):40156-40166).

Immunogen

CD47 purified from human spleen cells (Liu, Y., et al. (2001). J. Biol. Chem. 276(43):40156-40166).
Epitope: extracellular domain

Application

Immunoprecipitation Analysis: A representative lot immunoprecipitated an increased amount of surface CD47 from human polymorphonuclear (PMN) neutrophils following fMLP stimulation and after transmigration (Parkos, C.A., et al. (1996). J. Cell Biol. 132(3):437-450).

Western Blotting Analysis: A representative lot detected CD47 distribution among human polymorphonuclear (PMN) neutrophil subcellular fractions. Majority of CD47 was found co-localized with CD11b in the CD11b and secondary granules in unstimulated PMN and upregulated plasma membrane localization was seen following fMLP stimulation (Parkos, C.A., et al. (1996). J. Cell Biol. 132(3):437-450).
Research Category
Inflammation & Immunology
This mouse monoclonal Anti-CD47, clone PF3.1 Antibody, Cat. No. MABF958 is validated for use in Flow Cytometry, Immunoprecipitation, and Western Blotting for the detection of CD47.

Quality

Evaluated by Flow Cytometry in human PBMCs.

Flow Cytometry Analysis: 0.2 µL of this antibody detected CD47 surface expression on the gated lymphocytes population among one million human PBMCs.

Target description

33.47/30.00/31.45/32.10 kDa (mature isoform OA3-323/OA3-293/OA3-305/OA3-312) calculated. ~60-65 kDa (glycosylated) and ~35 kDa (deglycosylated) reported (Parkos, C.A., et al. (1996). J. Cell Biol. 132(3):437-450).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service