Skip to Content
MilliporeSigma
All Photos(3)

Documents

MABF945

Sigma-Aldrich

Anti-TAP2 Antibody, clone 429.4

clone 429.4, from mouse

Synonym(s):

Antigen peptide transporter 2, APT2, ATP-binding cassette sub-family B member 3, Peptide supply factor 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF-2, Really interesting new gene 11 prot

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

429.4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TAP2(6891)

General description

Antigen peptide transporter 2 (UniProt Q03519; also known as APT2, ATP-binding cassette sub-family B member 3, Peptide supply factor 2, Peptide transporter involved in antigen processing 2, Peptide transporter PSF2, Peptide transporter TAP2, PSF-2, Really interesting new gene 11 protein) is encoded by the TAP2 (also known as ABCB3, PSF2, RING11, Y1) gene (Gene ID 6891) in human. TAP2 (ABCB3) belongs to the subfamily B of ATP-binding cassette (ABC) transporters, members of which also include TAP1 (ABCB2) and multidrug resistance protein 1 (MDR1, ABCB1, P-gp). TAP is a heterodimeric peptide transporter composed of TAP1 and TAP2 that dimerize via their nucleotide-binding domain (NBD; a.a. 503-510) in a head-to-tail fashion. TAP preferentially mediates the transport of 8- to 16-a.a. peptides into the ER in a process powered by ATP-binding and hydrolysis. TAP1 and TAP2 harbor a specialized N-terminal transmembrane domain (TMD0) that is followed by the TAP half-transporter core module composed of one TMD and one NBD. TMD0 is composed of a 4-TM bundle that serves as the interaction platform for the assembly of the peptide-loading complex (PLC).

Specificity

Clone 429.4 was subcloned from the original 429.3 hybridoma reported by van Endert, P.M., et al. (1994). Immunity. 1(6):491-500. The epitope lies within TAP2 ABC transporter domain. Clone 429 detected TAP2 in HeLa, but not in the ST-EMO B-lymphoblastoid cell line derived from a patient with homozygous TAP2 deletion (TAP2-/-) (Courtesy of Peter van Endert, Ph.D., Institut Necker Enfants Malades (INEM), Centre De Medecine Moleculaire, Paris, France).

Immunogen

Baculovirus-expressed recombinant human TAP2 C-terminal fragment (van Endert, P.M., et al. (1994). Immunity. 1(6):491-500).

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TAP2 in U2OS human osteosarcoma cells.

Immunocytochemistry Analysis: A 1:400 dilution from a representative lot detected TAP2 in HeLa, but not in the ST-EMO B-lymphoblastoid cell line derived from a patient with homozygous TAP2 deletion (TAP2-/-) (Courtesy of Peter van Endert, Ph.D., Institut Necker Enfants Malades (INEM), Centre De Medecine Moleculaire, Paris, France).
Research Category
Inflammation & Immunology
This mouse monoclonal Anti-TAP2 Antibody, clone 429.4, Cat. No. MABF945 is validated for use in Immunocytochemistry and Western Blotting for the detection of TAP2.

Quality

Evaluated by Western Blotting in Raji cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected TAP2 in 10 µg of Raji cell lysate.

Target description

~70 kDa observed. 75.66/72.00 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radhika Gudi et al.
The Journal of cell biology, 193(4), 711-725 (2011-05-18)
Centrobin is a daughter centriole protein that is essential for centrosome duplication. However, the molecular mechanism by which centrobin functions during centriole duplication remains undefined. In this study, we show that centrobin interacts with tubulin directly, and centrobin-tubulin interaction is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service