Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABF918

Sigma-Aldrich

Anti-EAT-2 Antibody, clone 8F12

clone 8F12, from rat

Synonym(s):

SH2 domain-containing protein 1B, EWS/FLI1-activated transcript 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8F12, monoclonal

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SH2D1B(117157)

General description

SH2 domain-containing protein 1B (UniProt: O35324; also known as EWS/FLI1-activated transcript 2, EAT-2) is encoded by the Sh2d1b (also known as Eat2, Eat2a, Sh2d1b1) gene in murine species. EAT-2 is a Src homology 2 (SH2) - domain-containing (aa 5-101) intracellular adaptor protein that is related to signaling lymphocytic activation molecule (SLAM) associated protein (SAP). EAT-2 and SAP are expressed in natural killer (NK) cells, and their combined expression appears to be essential for NK cells to kill abnormal hematopoietic cells. NK cells lacking either SAP or EAT-2 display partial defects in NK cell-mediated cytotoxicity toward abnormal hematopoietic cells and more severe defects are observed when NK cells lacking both adaptors. It plays a role in regulation of effector functions of natural killer (NK) cells by controlling signal transduction through CD244/2B4. It is proposed to act as an inhibitor of CD244-mediated NK cell function including cytotoxicity and interferon-gamma production. It is reported to repress natural cytotoxicity and interferon secretion by a mechanism involving tyrosine phosphorylation of its C-terminus. It binds to the phosphorylated receptors CD84, SLAMF1, LY9 and CD244 and does not bind to non-phosphorylated SLAMF1. Mice lacking EAT-2 are reported to have enhanced NK cell activation responses. EAT-2 is expressed at all stages of maturation in NK cells and in macrophages and dendritic cells. EAT-2 is also expressed in spleen, thymus, lung, kidney, heart, colon and small bowel. (Ref.: Luis-Alberto Pérez-Quintero, L-A., et al. (2014). J. Exp. Med. 211(4); 727-742).

Specificity

Clone 8F12 detects EWS/FLI1-activated transcript 2 (EAT-2) in human and murine cells.

Immunogen

GST-tagged full length recombinant murine SH2 domain-containing protein 1B (EAT-2).

Application

Anti-EAT-2, clone 8F12, Cat. No. MABF918, is a rat monoclonal antibody that detects SH2 domain-containing protein 1B (EAT-2) and has been tested for use in Western Blotting and Immunoprecipitation.
Immunoprecipitation Analysis: A representative lot immunoprecipitated EAT-2 (Perez-Quintero, L.A., et. al. (2014). J Exp Med. 211(4):727-42).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected EAT-2 in YT-S transfectants expressing mouse EAT-2 and YT-S wild type cell lysates (Courtesy of Ming-Chao Zhong, Research Associate for Dr. Andre Veillette, Laboratory of Molecular Oncology, Clinical Research Institute of Montreal, Montreal, Quebec, Canada.).

Western Blotting Analysis: A representative lot detected EAT-2 in Western Blotting applications (Cruz-Munoz, M.E., et. al. (2009). Nat Immunol. 10(3):297-305; Guo, H., et. al. (2009). Mol Cell Biol. 35(1):41-51; Perez-Quintero, L.A., et. al. (2014). J Exp Med. 211(4):727-42.
Research Category
Signaling

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be rat IgG2a.

Target description

15.26 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service