Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF464

Sigma-Aldrich

Anti-Foxp3 Antibody (mouse), PE, clone 3G3

clone 3G3, 0.2 mg/mL, from mouse

Synonym(s):

Forkhead box protein P3, Scurfin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

3G3, monoclonal

species reactivity

mouse

concentration

0.2 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Foxp3(20371)

Related Categories

General description

The 3G3 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is expressed at constitutively high levels in Treg cells, which are further identified as being CD4+ CD25+. In resting conventional T cells (CD4+ CD25-) Foxp3 expression is restricted, and upon TCR activation is expressed only transiently and in a small proportion of cells. However, the growth factor TGF-beta has been shown to induce expression of Foxp3 in naïve T cells, driving their development into Foxp3+ Tregs, which are called “induced” or “adaptive” Tregs. These cells are phenotypically similar to so-called “natural” Tregs (CD4+ CD25high Foxp3+) which originate in the thymus and comprise the majority of Treg cells. Tregs are critical for maintaining peripheral tolerance and are implicated in the development of autoimmunity.It is important to review the literature in choosing an antibody for the Foxp3 antigen in flow cytometry, as the potential for high background or non-specific staining may be observed. The 3G3 antibody may be used for intracellular detection of Foxp3 in cells from mouse and Rhesus macaque.
This mouse monoclonal antibody Anti-Foxp3 (mouse), PE & clone 3G3 validated for use in Flow Cytometry for the detection of Foxp3.

Application

This Anti-Foxp3 Antibody,clone 3G3, PE conjugated, is validated for usein Flow Cytometry for the detection of Foxp3.

Quality

The monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 µg of this antibody detected Foxp3 in one million C57Bl/6 splenocytes.

Physical form

PE conjugated

Purifiedby Size Exclusion Chromatography

PurifiedMouse Monoclonal Antibody in buffer containing 10 mM NaH2PO4, 150 mM NaCl, 0.1%gelatin, pH 7.2, and up to 0.1% sodium azide

Storage and Stability

It isrecommended to store the product undiluted at 2-8°C and protected fromprolonged exposure to light. Do not freeze.

Disclaimer

Unless otherwise stated in our catalog or other companydocumentation accompanying the product(s), our products are intended forresearch use only and are not to be used for any other purpose, which includesbut is not limited to, unauthorized commercial uses, in vitro diagnostic uses,ex vivo or in vivo therapeutic uses or any type of consumption or applicationto humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service