Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABF3182

Sigma-Aldrich

Anti-Flt3L Antibody, clone 11C9.10.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

11C9.10.1, monoclonal

mol wt

calculated mol wt 26.42 kDa
observed mol wt ~27 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... FLT3LG(2323)

General description

Fms-related tyrosine kinase 3 ligand (UniProt: P49771; also known as Flt3 ligand, Flt3L, SL cytokine) is encoded by the FLT3LG gene (Gene ID: 2323) in human. Flt3 ligand is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-26), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 27-184), a transmembrane domain (aa 185-205), and a cytoplasmic domain (aa 206-235). Flt3-ligand is an important regulator of hematopoiesis that synergizes with a number of colony-stimulating factors and interleukins. It is expressed on myeloid, lymphoid, and dendritic cell progenitors. Expression of its receptor is restricted only to hematopoietic progenitors, early B cells and myeloid cells. Binding of Flt3-ligand to its receptor (Flt3) leads to receptor homodimerization and subsequent conformational changes that result in phosphorylation of its tyrosine kinase domains. This leads to rapid activation of receptor, its internalization, downstream actions, and degradation. Administration of Flt-3 ligand to mice is shown to cause a significant expansion of both CD8 + and CD8 dendritic cells at multiple sites, including spleen, lymph nodes, thymus, liver, lung, and bone marrow. Three isoforms of Flt3 have been described that are produced by alternative splicing. Isoform one is a single-pass type I membrane protein, isoform 2 is a homodimeric secreted or extracellular protein, and isoform 3 lacks the first 82 amino acids. (Ref.: Tsapogas, P., et al. (2017). Int. J. Mol. Sci. 18(6): 1115).

Specificity

Clone 11C9.10.1 is a mouse monoclonal antibody that detects Fms-related tyrosine kinase 3 ligand (Flt3L). It targets an epitope within the extracellular domain.

Immunogen

His-tagged recombinant fragment corresponding to the 158 amino acids from the N-terminal, extracellular domain of human Fms-related tyrosine kinase 3 ligand (Flt3L).

Application

Quality Control Testing

Evaluated by Western Blotting with recombinant Flt3L protein.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected human recombinant Flt3L.

Tested Applications

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Flt3L in HeLa and A431 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Flt3L, clone 11C9.10.1, Cat. No. MABF3182, is mouse monoclonal antibody that detects Flt3L and is tested for use in Immunocytochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service