Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABF3171

Sigma-Aldrich

Anti-CXCR7 Antibody, clone 10D1

Synonym(s):

Atypical chemokine receptor 3, C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, Chemokine orphan receptor 1, G-protein coupled receptor 159, G-protein coupled receptor RDC1 homolog, RDC-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10D1, monoclonal

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
inhibition assay: suitable

isotype

IgG2a

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

Gene Information

human ... ACKR3(57007)

Specificity

Clone 10D1 is a mouse monoclonal antibody that detects CXCR7. It targets an epitope within the extracellular domain.

Immunogen

Mouse L1.2 cells transfected with human CXCR7 and stimulated for 20 h with butyric acid (5 mM).

Application

Quality Control Testing

Evaluated by Flow Cytometry in Raji cells.

Flow Cytometry Analysis: 1 µg of this antibody detected CXCR7 in one million Raji cells.

Tested Applications

Inhibition Assay: A representative lot inhibited the binding to the principal ligand CXCL12 to CXCR7. (Patent ID: WO2010141986A1).

Flow Cytometry Analysis: A representative lot detected CXCR7 in Flow Cytometry applications (Patent ID: WO2010141986A1).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Atypical chemokine receptor 3 (UniProt: P25106; also known as C-X-C chemokine receptor type 7, CXC-R7, CXCR-7, Chemokine orphan receptor 1, G-protein coupled receptor 159, G-protein coupled receptor RDC1 homolog, RDC-1) is encoded by the ACKR3 (also known as CMKOR1, CXCR7, GPR159, RDC1) gene (Gene ID: 57007) in human. CXCR7 is a multi-pass membrane protein with four extracellular domains, seven transmembrane domains, and four cytoplasmic domains. It predominantly localized to endocytic vesicles and upon stimulation by the ligand internalizes via clathrin-coated pits in a β-arrestin-dependent manner. Once internalized, the ligand dissociates from the receptor and is targeted for degradation while the receptor is recycled back to the cell membrane. CXCR7 is an atypical chemokine receptor that controls chemokine levels and localization via high-affinity chemokine binding that is uncoupled from classic ligand-driven signal transduction cascades, resulting instead in chemokine sequestration, degradation, or transcytosis. It is also reported to act as a chemokine-scavenging receptor or chemokine decoy receptor. It acts a as a receptor for chemokines CXCL11 and CXCL12/SDF1. Chemokine binding to CXCR7 does not activate G-protein-mediated signal transduction but instead induces β-arrestin recruitment, leading to ligand internalization and activation of MAPK signaling pathway. In glioma cells, it is reported to transduce signals via MEK/ERK pathway, mediating resistance to apoptosis and promote cell growth and survival. In normal hematopoietic progenitors it is not involved in cell migration, adhesion, or proliferation. However, in malignant cells, its activation by CXCL11 leads to ERK1/2 phosphorylation that causes enhanced cell adhesion and migration. (Ref.: Wang, C., et al. (2018). Front. Pharmacol. 9; 641; Rajagopal, S., et al. (2010). Proc.Natl. Acad.Sci. USA. 107(2); 628-632; Levoye, A., et al. (2009). Blood. 113(24); 6085-6093; Zabel, BA., et al. (2009). J. Immunol. 183(5); 3204-3211).

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service