Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF3169

Sigma-Aldrich

Anti-P-selectin/CD62P Antibody, clone WAPS12.2

Synonym(s):

CD62 antigen-like family member P, CD62P, GMP-140, Granule membrane protein 140, LECAM3, Leukocyte-endothelial cell adhesion molecule 3, PADGEM, Platelet activation dependent granule-external membrane protein

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$232.00
100 μG
$583.00

$232.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
$232.00
100 μG
$583.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$232.00


Available to ship onApril 09, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

WAPS12.2, monoclonal

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
inhibition assay: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

-10 to -25°C

Gene Information

human ... SELP(6403)

Specificity

Clone WAPS12.2 is a mouse monoclonal antibody that detects P-selectin/CD62P.

Immunogen

L1/2m stably transfected with a human P-selectin cDNA construct.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human bone marrow tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:25 dilution of this antibody detected P-selectin/CD62P in Human bone marrow tissue sections.

Tested Applications

Inhibition Assay: A representative lot Inhibited the rolling of / T cells on thrombin-activated platelets. (Jutila, M.A., et al. (1994). J Immunol. 153(9):3917-28).

Flow Cytometry Analysis: A representative lot detected P-selectin/CD62P in Flow Cytometry applications (Jutila, M.A., et al. (1994). J Immunol.153(9):3917-28).

Immunohistochemistry Applications: A representative lot detected P-selectin (CD62P) in Immunohistochemistry applications (Auvinen, K., et al. (2014). Immunology. 143(3):406-15).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

P-selectin (UniProt: P16109; also known as CD62 antigen-like family member P, Granule membrane protein 140, GMP-140, Leukocyte-endothelial cell adhesion molecule 3, LECAM3, Platelet activation dependent granule-external membrane protein, PADGEM, CD62P) is encoded by the SELP (also known as GMRP, GRMP) gene (Gene ID: 6403) in human. CD62P is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-41), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 42-771), a transmembrane domain (aa 772-795), and a cytoplasmic domain (aa 796-830). It serves as a calcium-dependent receptor for myeloid cells that binds to carbohydrates on neutrophils and monocytes. The ligand recognized by CD62P is reported to be sialyl-Lewis X. CD62P mediates the interaction of activated endothelial cells or platelets with leukocytes and facilitates rapid rolling of leukocyte rolling over vascular surfaces during the initial steps in inflammation through interaction with selectin P ligand (CD162). Within a few minutes following stimulation with thrombin, histamine, or phorbol 12-myristate 13-acetate (PMA), CD62P is translocated to the cell surface. (Ref.: Somers, WS., et al. (2000). Cell. 103(3); 467-479; Jutila, MA., et al. (1994). J. Immunol. 153(9); 3917-3928).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service