Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABF3160

Sigma-Aldrich

Anti-N-Cadherin Antibody, clone 8C11

Synonym(s):

Cadherin-2;CDw325;Neural Cadherin;CD325

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$200.00
100 μL
$504.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$200.00
100 μL
$504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8C11, monoclonal

mol wt

calculated mol wt 99.81 and 97.04 kDa
observed mol wt ~105 and 130 kDa

purified by

using protein G

species reactivity

hamster, human

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CHD2(1000)

General description

Cadherin-2 (UniProt: P19022; also known as CDw325, Neural cadherin, N-cadherin, CD325) is encoded by the CDH2 (also known as CDNH, NCAD) gene (Gene ID: 1000) in human. N-Cadherin is a single-pass type I membrane glycoprotein protein that plays a role in calcium-dependent cell adhesion. Its expression is observed in cells of the nervous tissue, vascular smooth muscle cells, and myocytes. N-cadherins preferentially interacts with themselves in a homophilic manner in connecting cells; may thus contribute to the sorting of heterogeneous cell types. N-cadherin are synthesized as precursor proteins with a signal peptide (aa 1-25) and a propeptide (aa 26-159) that are proteolytically cleaved to generate functional, mature protein. Newly synthesized proN-cadherin (aa 1-906) is phosphorylated and proteolytically processed prior to transport to the plasma membrane. The mature N-Cadherin contains an extracellular domain (aa 160-724) with five homologous repeats that mediates adhesion, a single pass transmembrane domain (aa 725-745), and a conserved cytoplasmic domain (aa 746-906) that interacts with catenins to link cadherins to the actin cytoskeleton. N-cadherin is well expressed in endothelial cells and plays an essential role in the maturation and stabilization of normal vessels. However, it also promotes tumor cell survival, migration and invasion, and a high level of its expression is often associated with poor prognosis. A 69 amino acid segment of EC-4 of N-cadherin is reported to be essential and sufficient to increase cell motility and promote epithelial to mesenchymal transition. (Ref.: Kim, JB., et al. (2000). J. Cell Biol. 151(6); 1193-1205; Navarro, P., et al. (1998). J. Cell Biol. 140(6); 1475-1484).

Specificity

Clone 8C11 is a mouse monoclonal antibody that detects N-Cadherin. It targets an epitope within the extracellular domain.

Immunogen

MBP-tagged recombinant fragment corresponding to 502 amino acids from the extracellular domain of human N-Cadherin.

Application

Quality Control Testing

Evaluated by Western Blotting in Human heart tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cadherin-2 in Human heart tissue lysate.

Tested applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected N-Cadherin in Human brain and cerebellum tissue extracts.

Western Blotting Analysis: A representative lot detected N-Cadherin in Western Blotting applications (Kim, J.B., et al. (2000). J Cell Biol. 151(6):1193-1206).

Flow Cytometry Analysis: 1 μg from a representative lot detected N-Cadherin in one million KG-1 cells.

Inhibition Assay: A representative lot inhibited motility of BT20 breast cancer cells (Kim, J.B., et al. (2000). J Cell Biol. 151(6):1193-1206).

Flow Cytometry Analysis: A representative lot detected N-Cadherin in Flow Cytometry applications (Wahlig, S., et al. (2020). Sci Rep. 10(1):1681).

Immunocytochemistry Analysis: A representative lot detected Cadherin-2 in Immunocytochemistry applications (Navarro, P., et al. (1998). J Cell Biol.;140(6):1475-1484).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-N-Cadherin, clone 8C11, Cat. No. MABF3160, is a mouse monoclonal antibody that detects N-Cadherin and is tested for use in Flow Cytometry, Immunocytochemistry, Inhibition Assay, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service