Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABF3047

Sigma-Aldrich

Anti-SCARB1 Antibody, clone AOB-1

Synonym(s):

Scavenger receptor class B member 1;SRB1;SR-BI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
clone:
AOB-1, monoclonal
application:
FACS
WB
species reactivity:
mouse
technique(s):
flow cytometry: suitable
western blot: suitable
citations:

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

AOB-1, monoclonal

mol wt

calculated mol wt 56.75 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Scarb1(20778)

General description

Scavenger receptor class B member 1 (UniProt: Q61009; also known as SRB1, SR-BI) is encoded by the Scarb1 (also known as Srb1) gene (Gene ID: 20778) in murine species. SR-BI (SCARB1) is a widely expressed, multi-pass membrane glycoprotein with two cytoplasmic domains (aa 1-11 and 462-509), one extracellular domain (aa 33-440) and two transmembrane domains (aa 12-32 and 441-461). It serves as a receptor for several different ligands such as phospholipids, cholesterol ester, lipoproteins, and phosphatidylserine. It is a key component in the reverse cholesterol transport pathway and thus plays an important role in lipid metabolism. It may also contribute to variations in plasma lipid levels during fasting. Two isoforms of SR-BI have been described that are produced by alternative splicing and both isoforms can serve as a receptor for HDL and mediate selective uptake of cholesteryl ether and HDL-dependent cholesterol efflux. It is reported to facilitate the flux of free and esterified cholesterol between the cell surface and ApoB-containing lipoproteins and modified lipoproteins, although less efficiently than HDL. SR-BI is also involved in phagocytosis of apoptotic cells, via its phosphatidylserine binding activity. Elevated expression of SR-BI has been reported in a wide variety of malignant cell lines such as breast, prostate, ovarian, pancreatic, nasopharyngeal, and colorectal cancers. In hepatocytes, it is reported to act as a receptor for hepatitis C virus and facilitates virus entry into cells. SR-B1 is also described as a silica receptor that through an extracellular -helix specifically recognizes amorphous and crystalline silica. However, it does not recognize titanium dioxide nanoparticles, latex nanoparticles, or monosodium urate crystals. It is involved in silica-induced pulmonary inflammation in murine models. Clone AOB-1 is shown to specifically bind to murine SR-BI on Jurkat.EcoR cells and blocks silica binding to these cells in a dose-dependent manner. (Ref.: Tsugita, M., et al. (2017). Cell Rep. 18; 1298-1311; Mooberry, LK., et al. (2016). Front. Pharmacol. 7; 466). Webb, NR., et al. (1998). J. Biol. Chem. 273(24); 15241-15248).

Specificity

Clone AOB-1 is a rat monoclonal antibody that detects Scavenger receptor class B member 1 (SCARB1).

Immunogen

NIH3T3 cells transfected with mouse SCARB1 (SR-B1).

Application

Quality Control Testing

Evaluated by Flow Cytometry in mSR-B1/Jurkat.EcoR cells.

Flow Cytometry Analysis (FC): 1.0 μg of this antibody detected SR-B1 in one million Jurkat.EcoR cells expressing murine SR-B1 (SCRAB1).

Tested Applications

Inhibition Assay; A representative lot of this antibody partially inhibited IL-1? secretion in silica stimulated bone marrow-derived macrophages (Tsugita, M., et al. (2017). Cell Rep.18(5); 1298-1311).

Flow Cytometry Analysis: A representative lot detected SCARB1 in Flow Cytometry application (Tsugita, M., et al. (2017). Cell Rep. 18(5);1298-1311).

Western Blotting Analysis: A representative lot detected SCARB1 in Western Blotting application (Tsugita, M., et al. (2017). Cell Rep. 18(5); 1298-1311).

Immunohistochemistry Applications: A representative lot detected SCARB1 in Immunohistochemistry applications (Tsugita, M., et al. (2017). Cell Rep.18(5);1298-1311).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SCARB1, clone AOB-1, Cat. No. MABF3047, is a rat monoclonal antibody that detects SCARB1 and is tested for use in Flow Cytometry, Immunohistochemistry, Inhibition Assay, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG1 in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service